5014iQ Beta Attenuation Monitor - FAQs
5014iQ Beta Attenuation Monitor - FAQs
View additional product information for 5014iQ Beta Attenuation Monitor - FAQs (5014IQ)
12 product FAQs found
What is the difference between the Thermo Scientific 5014iQ Beta Attenuation Monitor and the Thermo Scientific 5030iQ SHARP Monitor?
Both instruments use beta attenuation for particulate mass measurement. The Thermo Scientific 5014iQ Beta Attenuation Monitor uses beta attenuation as its primary measurement method, while the Thermo Scientific 5030iQ SHARP Monitor combines beta attenuation with light-scattering nephelometry. Consider the 5014iQ for applications centered on continuous beta-based mass measurement and the 5030iQ SHARP Monitor when rapid optical response combined with beta correction is important.
Can the Thermo Scientific 5014iQ Beta Attenuation Monitor be used for outdoor ambient air monitoring?
Yes. The Thermo Scientific 5014iQ Beta Attenuation Monitor can be used for outdoor ambient air monitoring when appropriately weather protected. An optional outdoor enclosure is available for installations requiring additional environmental protection. Users should follow the specified installation and operating requirements for the selected configuration.
Can the Thermo Scientific 5014iQ Beta Attenuation Monitor be accessed remotely?
Yes. The Thermo Scientific 5014iQ Beta Attenuation Monitor includes Ethernet connectivity and remote data access for integration into air quality monitoring networks. Supported communications protocols include MODBUS TCP/IP, streaming data, and NTP.
Can the Thermo Scientific 5014iQ Beta Attenuation Monitor operate unattended?
Yes. The Thermo Scientific 5014iQ Beta Attenuation Monitor is designed for long-term unattended operation. The filter tape can advance automatically based on operating parameters such as accumulated mass and sample flow, helping reduce routine operator intervention.
How frequently does the Thermo Scientific 5014iQ Beta Attenuation Monitor output particulate measurement data?
The Thermo Scientific 5014iQ Beta Attenuation Monitor outputs data every second. For U.S. EPA equivalent method operation, however, the PM-10 and PM-2.5 designations specify 24-hour average measurements, so regulatory reporting, averaging, completeness, and validation should follow applicable EPA and monitoring-agency requirements.
How does the Thermo Scientific 5014iQ Beta Attenuation Monitor handle high particulate loading or smoke events?
The Thermo Scientific 5014iQ Beta Attenuation Monitor tracks collected particulate mass and sample flow and can advance the filter tape to a clean sampling area when configured thresholds are reached. Because the instrument measures mass by beta attenuation rather than optical response alone, particle color or optical properties do not directly affect the beta measurement.
How does the Thermo Scientific 5014iQ Beta Attenuation Monitor manage changing humidity conditions?
The Thermo Scientific 5014iQ Beta Attenuation Monitor monitors sample conditions and applies heat only when needed to help reduce moisture-related measurement effects. This dynamic heating approach also helps preserve volatile aerosol components during measurement.
Which inlet configuration should I use with the Thermo Scientific 5014iQ Beta Attenuation Monitor?
Select the inlet configuration based on whether your application requires PM-2.5, PM-10, PM-1, or TSP monitoring. For U.S. EPA PM-10 or PM-2.5 regulatory monitoring, use the configuration specified in the applicable equivalent method designation and operating manual.
Can the Thermo Scientific 5014iQ Beta Attenuation Monitor be used for U.S. EPA regulatory PM monitoring?
Yes, when configured and operated as specified in the applicable U.S. EPA designation. The Thermo Scientific 5014iQ Beta Attenuation Monitor is included under automated equivalent method designations EQPM-1102-150 for PM-10 and EQPM-0609-183 for PM-2.5. These designations apply only to the specified configurations and operating conditions; PM-1 and TSP measurement capability is not covered by these designations. Users should follow the current EPA designation and applicable monitoring-agency requirements.
What are the advantages of continuous sampling with the Thermo Scientific 5014iQ Beta Attenuation Monitor?
The Thermo Scientific 5014iQ Beta Attenuation Monitor uses continuous, non-stepwise sampling to support continuous particulate measurement and help mitigate particle loss associated with interrupted sampling. Its automated filter-tape system also supports long-term unattended operation.
How does the Thermo Scientific 5014iQ Beta Attenuation Monitor measure particulate matter?
The Thermo Scientific 5014iQ Beta Attenuation Monitor collects particulate matter on a filter tape and uses beta attenuation to determine the deposited mass. The instrument also measures sample volume, allowing it to calculate ambient particulate mass concentration.
What does the Thermo Scientific 5014iQ Beta Attenuation Monitor measure?
The Thermo Scientific 5014iQ Beta Attenuation Monitor measures particulate matter concentrations in ambient air using beta attenuation. With the appropriate inlet configuration, it supports PM-2.5, PM-10, PM-1, or total suspended particulate (TSP) monitoring.