Where can I find the manual for the BindIt Software?
I cannot connect my KingFisher Flex instrument to the BindIt software. How do I troubleshoot this issue?
To determine the cause of the connection issue, we recommend checking the following:
1. Check software settings:
• Make sure the instrument selected is KingFisher Flex.
• Make sure the instrument serial number was input with the dash included and no spaces (ex, "711-xxxx").
• Make sure the communication tab was set to "USB" and not "COM".
2. Check the instrument and computer:
• The KingFisher Flex instrument screen (GUI) should be set to the "Factory Protocols" tab (blue tab) for best results.
• Make sure the port function on the Maintenance Tab (last tab) is checked for "USB".
• Try using different USB cables.
• Try a different USB port on the computer.
• Check the Windows version you are using.
• Reboot the KingFisher Flex instrument and the computer.
• Make sure the driver was installed properly. This may require the help of your local IT team.
If the connection issue persists, please contact our Technical Support team at techsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
What Uninterruptible Power Supply (UPS) do you recommend I use with the KingFisher Flex instrument?
You can use any brand of UPS as long as it is compatible with the following KingFisher Flex instrument electrical specifications:
• Mains power supply: 100–240 Vac, 50/60 Hz, nominal automatic voltage detection
• Power consumption: 175 VA max.; 55 VA standby
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
I am getting the following error message on the KingFisher Flex instrument: error message 12. What can I do to fix this error?
The error message 12 error usually means there is something out of place on the KingFisher Flex instrument. We recommend the following to resolve the issue:
1. Check the heat block and make sure that it is not lifted up out of the base of the unit. If it is out of the base, check to see if there is anything underneath it that is preventing it from going down. If there is nothing under the heat block, it may have gotten stuck in the up position. This will require a service call.
2. Check that the tip shield is off to the right and out of the way from underneath the magnetic head. If it is stuck in the incorrect position, this may require a service call.
3. If the turntable was moved and it clicked out of place, the rectangular footprint cannot be centered below the head. Turn off the instrument and turn it back on. The instrument should re-align itself.
If the error continues to display, please contact techsupport@thermofisher.com for assistance.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
How do I use BindIt software 4.0 to load a script from my computer to the KingFisher Flex or KingFisher Duo/Duo Prime instrument?
Please follow these steps to load a script from your computer to the KingFisher Flex or KingFisher Duo/Duo Prime instrument:
1. Use the USB cable to connect the PC to the instrument.
2. Turn on the instrument.
3. Open the BindIt software and click the Connect button.
4. Select "Find New Instrument." BindIt will automatically connect to the instrument and the instrument screen will say "Computer Control".
5. Click the "Transfer" Button.
6. Click the "Upload" button.
7. Browse to find the file and double-click it.
8. In the new window that opens:
a. KingFisher Flex: you will be prompted to enter a new name – this is not necessary if the name only contains letters, digits, dash or underscore.
• If the name contains spaces, apostrophes or any other character, you should enter a new name containing only letters, digits, dash (-) or underscore (_).
• Click the "Category" pulldown to select the instrument folder you want to load the script to.
• Click OK.
b. KingFisher Duo/Duo Prime: The KingFisher Duo and Duo Prime instruments accept all characters in the filename.
• Click the "Category" pulldown to select the instrument folder you want to load the script to.
• Click OK.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
I am using the KingFisher Flex instrument and see that there is a warning in the user guide that people wearing a pacemaker should not come into close contact with the magnet. What distance is safe for people with pacemakers to be at?
We recommend that people with pacemakers consult with their doctor before working with the KingFisher instruments. For the 24 Deep-Well Head, the magnetic force is 3,500 gauss. For the 96 Deep-Well Head, the magnetic force is 1,700 gauss. For the other magnetic heads, the magnetic force is lower.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
Do the KingFisher Flex and KingFisher Duo instruments come with pre-loaded scripts?
The KingFisher Flex and KingFisher Duo instruments no longer come with pre-installed scripts. If your instrument begins with the following serial numbers, it will not have pre-loaded scripts:
• KingFisher Flex serial number 711-1951 or greater
• KingFisher Duo Prime serial number 706-1366 or greater
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
How can I calibrate the heating block for the KingFisher Flex instrument if it is off by 1 degree Celsius or more?
The accuracy of the KingFisher Flex heating block is ± 1°C for up to 80°C, and ± 2°C for up to 115°C (see page 72 of the
manual). If there is an issue with the accuracy of the heating block, you can contact to schedule a service call. Calibration of the heating block can only be performed by one of our engineers.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
Can I transfer a script protocol from my KingFisher Flex instrument back to the computer so that I can edit the protocol?
No, scripts cannot be transferred from the KingFisher Flex instrument to the computer for editing. It is possible to transfer a backup copy of the script from the KingFisher Flex instrument to the computer, but you will not be able to edit it. You may use this backup file to write a new script. In general, we recommend storing protocols in the BindIt software on the computer so the protocols can be edited there.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
Which plastic consumables can I use with the KingFisher Flex 24 Deep-Well Head?
Do I receive training when I purchase the KingFisher Flex or KingFisher Duo Prime instruments?
No training is provided with the purchase of a KingFisher Flex or KingFisher Duo Prime instrument, but you can purchase training separately. We offer onsite half-day and full-day trainings for the KingFisher Flex and KingFisher Duo Prime instruments, depending on your needs. Please contact your local instrument sales representative to obtain a quote. The training will be led by your regional Field Application Scientist.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
I am using a purification kit from a different company. Can you help me write a script to run on the KingFisher Flex instrument?
We cannot write a script for a purification kit for another company, but we can provide you with a tutorial on how to use the BindIt software. We also recommend checking with the manufacturer of the purification kit you are using, since some companies may already have scripts for their products to be used on multiple instruments.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
Where can I find a script for processing a large volume of blood using the MagMAX Multi-Sample Ultra kit (Cat. No. A25597) with the KingFisher Flex or KingFisher Duo Prime instruments?
Scripts for processing large volumes of whole blood (up to 1 mL) can be found on the
MagMAX Multi-Sample Ultra kit product page under the Product Literature section at the bottom of the page. There are two scripts available:
• KingFisher DUO file: A25597_Large_Volume_Blood.bdz
• KingFisher Flex file: A25597_Large_Volume_Blood.bdz
*Use the page scrolling function to see the complete list of product literature for this product.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
What kind of training do you offer for the KingFisher Flex instrument and how much does it cost?
We offer onsite half-day and full-day trainings for the KingFisher Flex instruments, depending on your needs.The training will be led by your regional Field Application Scientist. You can contact your local sales representative to obtain a quote for the training cost.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
What should I do if the tip comb does not load correctly onto my Thermo Scientific KingFisher system?
If the tip does not load correctly onto a Thermo Scientific KingFisher magnetic head, we recommend that you try manually flexing the tips both lengthwise and widthwise to level the tip comb. A video of flexing the tip comb can be found here: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/automated-purification-extraction/kingfisher-systems/resources.html. The video is under "KingFisher Flex" and titled "How to install the magnet head and check the tip comb." Flexing of the tip comb is demonstrated at timepoint 1:35. If that does not work, try a different box or lot of tips. This applies to all KingFisher instruments.
Can I use the KingFisher instruments for plasmid purification?
We recommend using the MagJet Plasmid DNA kit (Cat. No. K2791) for plasmid purification. Scripts for using the MagJet Plasmid DNA kit with the KingFisher Flex and KingFisher Duo Prime Purification Systems are available for download on the kit product page (https://www.thermofisher.com/order/catalog/product/K2791). The ChargeSwitch Plasmid ER Mini Kit (Cat. No. CS10100) is also an option for plasmid purification. There is no script currently available for using ChargeSwitch Plasmid ER Mini Kit, but you can create a custom protocol for use with the KingFisher Flex and KingFisher Duo Prime Purification Systems.
Are the magnets on the KingFisher Flex Purification System magnetic head strong enough to disturb nearby laboratory equipment?
We do not recommend placing the KingFisher Flex Purification System in close proximity to magnetic tapes, computer discs or other magnetic storage systems, such as credit cards, as these can be damaged by the strong magnetic field of the KingFisher Flex magnetic heads. Do not hold the KingFisher Flex magnetic heads close to a PC display, since this may cause damage to the display. Do not use metal tools when handling KingFisher Flex magnetic heads. People wearing a pacemaker or metallic prostheses should also not use this product as these items could be affected or damaged by close contact with a strong magnetic field.
How should I clean the cover on the KingFisher Flex instrument?
We recommend cleaning the KingFisher Flex instrument cover with water containing a mild soap or a slightly acidic, neutral or slightly alkaline detergent. You can use up to a 3.5% Sodium Hypochlorite concentration. We do not recommend using phenol or chloroform to clean the cover.
What material is the KingFisher Flex Purification System cover made of?
The KingFisher Flex Purification System cover is made of polyethylene terephthalate (PET).
How does the KingFisher Flex Purification heating block work?
The KingFisher Flex Purification System heating block is located inside the instrument and can be used automatically during the protocol. All KingFisher Flex plates can be heated using specially designed, interchangeable heating blocks. Any number of heating steps can be added to a protocol. When a protocol enters a heating step, the plate is automatically moved to the dedicated heating position for heating. Afterwards, the protocol continues automatically to the next step.
What is the advantage of using the KingFisher Flex instrument with the 24 Deep-Well magnetic head?
The KingFisher Flex instrument with the 24 Deep-Well magnetic head can process larger volumes for each sample (up to 5mL total volume) compared to the KingFisher Flex instrument with the 96 Deep-Well magnetic head.
Can I use the same tips and plates for the KingFisher Flex instrument with the 96-well magnetic head and the MagMAX Express-96 instrument magnetic head?
Yes, the KingFisher Flex instrument 96-well magnetic head and the MagMAX Express-96 instrument magnetic head can use the same tip combs and KingFisher plates even though the magnetic heads are different.
Where can I find scripts for the MagMAX Express-96 instrument?
The MagMAX Express-96 protocol scripts can be found on the MagMAX Express protocol page (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/total-rna-extraction/magmax-technology/magmax-particle-processors/magmax-instrument-protocols-scripts.html). If the specific script you are looking for is not located here or on the product page for the MagMAX kit, please contact Technical Support at techsupport@thermofisher.com.
Which software should I use for transferring scripts to my MagMAX Express-96 or KingFisher Flex instrument?
We recommend using the BindIt Software to transfer scripts to the MagMAX Express-96 and KingFisher Flex instruments. The BindIt Software should be used to transfer scripts for all of the Applied Biosystems MagMAX Express and Thermo Scientific KingFisher instruments.
Can I re-start a script that has stopped mid-way on the KingFisher Flex/KingFisher Duo Prime instrument?
The only way is to do this is by rewriting the script from where you want it to start, or start the run all over again.
I have a BindiT protocol installed on the KingFisher Flex/DuoPrime instrument and would like to edit it. How do I do it?
There is no way to edit a BindiT protocol that has been installed on the KingFisher Flex/KingFisher DuoPrime instrument. The only way to edit the protocol is by saving the original .bdz file to a PC and then editing it in BindiT software.
I cannot transfer files to the KingFisher Flex instrument using the BindIT software. What should I do?
The KingFisher Flex instrument needs to be connected to the computer/laptop and recognized by it. The lower bottom screen should say which instrument it is connected to. If it says anything about simulator, it is not connected to a real instrument.
You need to add a new instrument to the database in order to recognize the instrument: Settings > Instrument Setup > New > select what kind of instrument it is from the drop-down menu (i.e., KingFisher 96/MagMAX Express 96), give the instrument a name, and enter the serial number (make sure to keep the - in the middle). Once the instrument is connected and recognized, you should be able to transfer files over.
I am getting an error message with the MagMAX Express/KingFisher Flex instrument. What should I do?
Turn the instrument on and off to see whether the error message disappears. Alternatively, do a dry run without any samples. If the issue could be consumable dependent try a new set/lot of tip combs. If the error message repeatedly occurs, please contact Technical Support (techsupport@thermofisher.com).
Can I make changes to a .kf2 file in the BindIt software?
A .kf2 file cannot be edited in the BindIt software, but it is possible to rewrite a .kf2 file as a .bdz file so that it can be edited. Please see this application note for instructions on how to convert a .kf2 file into a .bdz file (http://www.thermoscientific.com/content/dam/tfs/LPG/LCD/LCD Documents/Application & Technical Notes/Liquid Handling and Dispensing/KingFisher-BindIT-TechNote-5189009.pdf). The speed settings are different in the two software (KingFisher software has 13 speed settings and BindIt software has 5 speed settings), so the application note has a conversion table for what speed to use in the BindIt software.
What is the difference between a .bdz file and a .kf2 file?
They are created by different software (BindIt and KingFisher software, respectively).
.kf2 files can only be used for the MagMAX Express instrument whereas .bdz files can be used for MagMAX Express and KingFisher Flex/Duo instruments (but scripts need to be specifically written for a particular instrument).
What is the difference between the BindIt software and old KingFisher 2.6 software?
The old KingFisher 2.6 software only works on Windows XP, whereas the BindIt software works on Windows XP, Windows 7, and Windows 8. The speed selections are also different. The old KingFisher 2.6 software featured 13 different speed setting whereas the BindIt software features 5 mixing speeds (slow, medium, fast, bottom, and half mix). The BindIt software can run KingFisher Software 2.6 files (.kf2) just as they are, but it cannot edit the .kf2 file. Please review our
Technical Note for instructions on how to convert a .kf2 file to a format that can be edited in the BindIt software. A .kf2 protocol thas has been converted into a BindIt protocol can still achieve excellent yield and purity of the extracted target. Minor optimization may be required due to the differences in mix speeds and bead collection parameters, or to minimize protocol times.
Where can I find the KingFisher Flex or KingFisher Duo Prime scripts for the animal health MagMAX kits?
The scripts for the MagMAX CORE Nucleic Acid Purification Kit can be found here (https://www.thermofisher.com/order/catalog/product/A32702). In order to obtain the .bdz scripts for the other animal health MagMAX kits (Cat. Nos. AM1835, AM1836, AM1840, 4462359), please email technical support at animalhealthandfoodsafety@lifetech.com.
What is the major difference between the KingFisher Flex and MagMAX Express-96 instruments?
- The KingFisher Flex instrument has the option of using a 24-Deep-Well head in addition to a 96-Deep-Well head.
- The KingFisher Flex instrument has a USB communications port, whereas the MagMAX Express-96 instrument only has a RS-232 port.
- The KingFisher Flex instrument uses snap-in heat blocks, while the heat blocks need to be screwed in for the MagMAX Express-96 instrument.
- Unlike the MagMAX Express-96 instrument, the KingFisher Flex instrument has a graphical user interface and expanded keypad (LCD screen).
Which automated magnetic particle processor should I use for animal health projects?
We recommend one of the following instruments:
- KingFisher Flex Purification System (Cat. No. 5400640)
- KingFisher Duo Prime Purification System (Cat. No. 5400110)
- MagMAX Express Magnetic Particle Processor (AM-Applied Markets Customer) (Cat. No. 4400075)
Can the KingFisher Flex heating blocks be purchased separately?
Yes, here are the catalog numbers for standalone heating blocks:
- Cat. No. 24075420: KingFisher Flex 96 KF Heating Block
- Cat. No. 24075430: KingFisher Flex 96 Deep-Well Heating Block
- Cat. No. 24075440: KingFisher Flex 24 Deep-Well Heating Block
Does the KingFisher Flex Magnetic Particle Processor with 24 Deep-Well Head come with a heating block?
Yes, it comes with the KingFisher Flex 24 Deep-Well Heating Block.
Does the MagMAX Cell-Free DNA Isolation Kit contain MagMAX magnetic beads?
No, the MagMAX Cell-Free DNA Isolation Kit contains the Dynabeads MyOne SILANE magnetic beads. For this reason we recommend using the DynaMag magnets (Cat. Nos. 12302D, 12321D).
What sample types can the MagMAX Cell-Free DNA Isolation Kit be used for?
The MagMAX Cell-Free DNA Isolation Kit has been designed to isolate cell-free DNA (cfDNA) from plasma, serum, and urine samples.
We are getting a lot of variation in cell-free DNA yield from donor to donor when using the MagMAX Cell-Free DNA Isolation Kit. Do you have any tips for how to improve that?
Levels of cell-free DNA in circulation can range from 1 ng/mL to 100 ng/mL of plasma or serum depending on the donor. For samples containing low levels of cell-free DNA, we recommend increasing the starting sample volume.
There is an abundance of gDNA in the eluate I obtained from the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319). Do you have any recommendations to prevent that from happening?
Yields from hemolytic plasma, lipemic plasma, or other compromised samples types vary greatly from donor to donor. We recommend processing these types of samples using the manual protocol.
Where can I find the scripts for automated use of the MagMAX mirVana Total RNA Isolation Kit?
You can find scripts for using MagMAX mirVana Total RNA Isolation Kit with the MagMAX Express-96, KingFisher Flex, or KingFisher Duo instrument, on the product page (http://www.thermofisher.com/order/catalog/product/A27828).
I see that the KingFisher 24 Deep Well Head has a processing volume of 5 mL. Does that mean that I can process up to 5 mL of my sample?
No, the processing volume refers to the maximum volume that can be held in the 24 Deep Well plates. This incorporates the volume of binding buffers and beads, so when scaling up reaction volumes, it is important to make sure that the total volume does not exceed 5 mL.
My KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?
No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original KingFisher Flex script would have to be created that starts off at the point where the run was stopped.
I have a 12-plate protocol but there are only 8 slots on the KingFisher Flex instrument turntable. What should I do with the remaining 4 plates?
Set up the protocol on the BindIt software. After the first 8 plates are done, the instrument will pause and instruct you to switch out the plates.
Is it always necessary to use the transport locks on the KingFisher Flex instrument?
The transport locks are only necessary when relocating the instrument.
Are the volumes of reagents in each well critical for a successful KingFisher Flex run?
We strongly recommend that you maintain the specified volumes within the defined limits to avoid spillover and to help ensure the best performance at the most efficient level.
What if the magnetic particles are attached to the tip combs after my KingFisher Flex run?
This happens sometimes but it will not affect the yield because the sample has been released from the particles.
What if the magnetic particles remain in the sample well during a KingFisher Flex run?
If the starting material is too viscous, the magnetic rods will not be able to collect the particles. Dilute the sample and check that the sample is properly homogenized/lysed. Adding low amounts of detergent will improve the collection of magnetic particles as well. Quickly centrifuge the plate to sediment the particles to the bottom of the plate.
What should I do if I forget to insert the tip combs into the tip comb holder for the KingFisher Flex instrument?
The protocol will not start without the tip combs inserted into the tip comb holder.
What should I do if the KingFisher Flex magnet sets are dirty?
Wipe the magnetic rods with a soft cloth or tissue paper soaked in a mild detergent solution, soap solution, or alcohol.
How long does it take the KingFisher Flex heater to warm up from room temperature to 80°C?
The heating block warms up at the rate of about 10°C per minute, so it will take about 6 minutes.
Can I concentrate samples during a KingFisher Flex run?
Both deep-well plates and KingFisher 96 plates can be used during the same run. Therefore, it is possible to start the processing by using larger volumes (in a deep-well plate) and elute the purified sample to a smaller volume (in a KingFisher 96 or MagMAX 96 standard plate).
How can the KingFisher Flex heads be changed?
This can be done by running the Change magnet protocol under the Maintenance menu using the up and down cursor keys.
Will the KingFisher Flex magnets get weaker? If so, how long can they be used?
The KingFisher Flex magnets are made of material that is very stable. The magnetic field will not get weaker. However, extreme mechanical force or heating can cause damage to the magnets.
Caution: It is very important to keep the KingFisher Flex heads away from each other and from other magnets at all times. Clashing together of the magnets may cause serious damage to the magnets.
How does the KingFisher Flex instrument connect to a computer?
KingFisher Flex uses a USB port, whereas the MagMAX Express- 96 requires a RS-232 port.
I have a custom protocol stored in my KingFisher Flex instrument. Can I edit this protocol and save it to my computer?
No. Protocols cannot be transferred back from the instrument to the computer. Protocols need to be previously stored in the BindIt or KingFisher software to make edits.
Does the BindIt software also work with the MagMAX Express-96 instrument?
Yes, the MagMAX Express-96 instrument is compatible with the BindIt software. A serial port-to-USB converter is required to connect the MagMAX Express-96 instrument to the computer.
If you are using BindIt software v3.3.1.7 you can use it to run existing .kf2 files. You will not be able to edit the .kf2 files in v3.3.1.7.
If you are using BindIt software v4.0 or later, you cannot run or edit existing .kf2 files. To get around this, you can convert the .kf2 files to the BindIt (.bdz) file format and then edit the .bdz file in the BindIt software. Please see the following application note for instructions on how to convert the .kf2 files: (http://www.thermoscientific.com/content/dam/tfs/LPG/LCD/LCD Documents/Application & Technical Notes/Liquid Handling and Dispensing/KingFisher-BindIT-TechNote-5189009.pdf). If you have already converted your .kf2 files to .bdz files, or you plan to convert your .kf2 file to .bdz files, we recommend using BindIt software v4.0 or higher.
Can I edit the manufactured KingFisher Flex protocols?
No, manufactured protocols are locked for editing, but they can be renamed using the save as' function and edited as a new version.
What Microsoft Windows version is the BindIt software compatible with?
BindIt software v3.3.1 is compatible with Windows XP (Professional with Service Pack 3), Windows Vista (Service Pack 2), Windows 7 (Service Pack 1), and Windows 8.
BindIT software v4 is compatible with Windows 7 with Service Pack 1 (32- or 64-bit), Windows 8.1 (32- or 64-bit), and Windows 10 (32- or 64-bit).
Can I use magnetic beads from other vendors with the KingFisher Flex instrument?
For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?
Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.
Can I scale up my reaction volume with the KingFisher Flex instrument?
Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.
Can I use plates from other manufacturers with the KingFisher Flex Magnetic Particle Processor?
No, we strongly recommend that you use the MagMAX plates or KingFisher plates since they were specifically designed to be used with KingFisher Flex tip combs to attain maximal performance. Plates from other manufacturers may not be compatible with the KingFisher Flex heating blocks. They may also cause unexpected problems, such as cross-contamination, due to the divergent well volume and bottom height of the plate.
How many magnetic head options are available for the KingFisher Flex Magnetic Particle Processor?
There are 4 magnetic head options available for the KingFisher Flex Magnetic Particle Processor:
- KingFisher Flex head for MagMAX Express-96 Deep Well plate
- KingFisher Flex head for KingFisher 24 Deep Well plate
- KingFisher Flex head for KingFisher 96 plate
- KingFisher Flex head for PCR plate, skirted
Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?
Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.