Dynabeads™ Streptavidin for In Vitro Transcription, 2 mL - FAQs
Dynabeads™ Streptavidin for In Vitro Transcription, 2 mL - FAQs
View additional product information for Dynabeads™ Streptavidin for In Vitro Transcription - FAQs (65005D, 65006D)
1 product FAQs found
What is the concentration of Dynabeads Streptavidin for In Vitro Transcription (Cat. Nos. 65005D, 65006D)?
The concentration of Dynabeads Streptavidin for In Vitro Transcription is 10 mg/mL, approximately 7-9 x 109 beads/mL.
Note: The numbers will be within this range, but vary from batch to batch.