Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Aldolase C Antibodies

          Proteintech

          Aldolase C Monoclonal Antibody (5H7A5)

          View all (11) Aldolase C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Aldolase C Monoclonal Antibody (5H7A5)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          Aldolase C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Aldolase C Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Aldolase C Antibody (66120-1-IG) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using Aldolase C antibody (Product # 66120-1-Ig) (Clone: 5H7A5) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Aldolase C Antibody in Immunocytochemistry (ICC/IF)
          Aldolase C Antibody in Immunocytochemistry (ICC/IF)
          Aldolase C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aldolase C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aldolase C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aldolase C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aldolase C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aldolase C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aldolase C Antibody in Western Blot (WB)
          Aldolase C Antibody in Western Blot (WB)
          Aldolase C Monoclonal Antibody (5H7A5)

          Product Details

          66120-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5H7A5

          Immunogen

          Aldolase C Fusion Protein Ag6779 (1-364 aa encoded by BC003613)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1.04 mg/mL

          Amount

          156 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MPHSYPALS AEQKKELSDI ALRIVAPGKG ILAADESVGS MAKRLSQIGV ENTEENRRLY RQVLFSADDR VKKCIGGVIF FHETLYQKDD NGVPFVRTIQ DKGIVVGIKV DKGVVPLAGT DGETTTQGLD GLSERCAQYK KDGADFAKWR CVLKISERTP SALAILENAN VLARYASICQ QNGIVPIVEP EILPDGDHDL KRCQYVTEKV LAAVYKALSD HHVYLEGTLL KPNMVTPGHA CPIKYTPEEI AMATVTALRR TVPPAVPGVT FLSGGQSEEE ASFNLNAINR CPLPRPWALT FSYGRALQAS ALNAWRGQRD NAGAATEEFI KRAEVNGLAA QGKYEGSGED GGAAAQSLYI ANHAY (1-364 aa encoded by BC003613)

          Target Information

          This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALDOC; aldolase 3; aldolase C, fructose-bisphosphate; Brain-type aldolase; EC 4.1.2.13; epididymis secretory sperm binding protein; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; Fructose-bisphosphate aldolase C; unnamed protein product

          View more View less

          Gene Aliases: ALDC; ALDOC

          View more View less

          UniProt ID: (Human) P09972

          View more View less

          Entrez Gene ID: (Human) 230

          View more View less

          Function(s)
          fructose-bisphosphate aldolase activity protein binding cytoskeletal protein binding lyase activity
          Process(es)
          fructose metabolic process gluconeogenesis glycolytic process fructose 1,6-bisphosphate metabolic process epithelial cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline