Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EDCNAMAFC AKMRSSKKTE VNLEAPEPGV EVIFYLSDRE PLRLGSGEYT AEELCIRAAQ ACRISPLCHN LFALYDENTK LWYAPNRTIT VDDKMSLRLH YRMRFYFTNW HGTNDNEQSV WRHSPKKQKN GYEKKKIPDA TPLLDASSLE YLFAQGQYDL VKCLAPIRDP KTEQDGHDIE NECLGMAVLA ISHYAMMKKM QLPELPKDIS YKRYIPETLN KSIRQRNLLT RMRINNVFKD FLKEFNNKTI CDSSVSTHDL KVKYLATLET LTKHYGAEIF ETSMLLISSE NEMNWFHSND GGNVLYYEVM VTGNLGIQWR HKPNVVSVEK EKNKLKRKKL ENKHKKDEEK NKIREEW (8-363 aa encoded by BC132729)
Janus kinase 1 (JAK1), is a protein tyrosine kinase that is involved in the interferon-alpha/beta and -gamma signal transduction pathways. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK1. JAK1 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to cis elements upstream of target gene promoters to regulate transcription. Upon ligand binding, JAK1 undergoes tyrosine phosphorylation and catalytic activation in an interdependent manner. Phosphorylation of tyrosine residues at positions 1022 and 1023 is believed to function in the activation of catalytic events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.10.2; JAK-1; Janus kinase 1; Janus protein tyrosine kinase 1; kinase Jak1; Tyrosine-protein kinase JAK1
Gene Aliases: AA960307; AIIDE; BAP004; C130039L05Rik; JAK1; JAK1A; JAK1B; JTK3
UniProt ID: (Human) P23458
Entrez Gene ID: (Human) 3716, (Rat) 84598, (Mouse) 16451
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support