Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GTDC1 Antibodies

          Proteintech

          GTDC1 Monoclonal Antibody (1C4C10)

          View all (8) GTDC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GTDC1 Monoclonal Antibody (1C4C10)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          GTDC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GTDC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GTDC1 Antibody (66578-1-IG) in ICC/IF

          Immunofluorescent analysis of ( 4% PFA) fixed U2OS cells using 66578-1-IG (GTDC1 antibody) at dilution of 1:100 and Alexa Fluor 488-Conjugated AffiniPure Goat Anti-Mouse IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GTDC1 Antibody in Immunocytochemistry (ICC/IF)
          GTDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GTDC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GTDC1 Antibody in Western Blot (WB)
          GTDC1 Monoclonal Antibody (1C4C10)

          Product Details

          66578-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Pig

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1C4C10

          Immunogen

          GTDC1 Fusion Protein Ag9934 (1-292 aa encoded by BC061699)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          150 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSILIIEAF YGGSHKQLVD LLQEELGDCV VYTLPAKKWH WRARTSALYF SQTIPISEHY RTLFASSVLN LTELAALRPD LGKLKKILYF HENQLIYPVK KCQERDFQYG YNQILSCLVA DVVVFNSVFN MESFLTSMGK FMKLIPDHRP KDLESIIRPK CQVIYFPIRF PDVSRFMPKH KTTHLKKMLG LKGNGGAVLS MALPFQPEQR DSEDLLKNFN SECDTHCGLD TARQEYLGNS LRQESDLKKS TSSDNSSSHH GENKQNLTVD PCDILGGVDN QQRLLHIVWP HRW (1-292 aa encoded by BC061699)

          Target Information

          The GTDC1 (Glycosyltransferase-Like Domain Containing 1) gene encodes a protein that is hypothesized to function as a glycosyltransferase. This gene has been implicated in several crucial biological processes, particularly in the nervous system. The GTDC1 cDNA is 2954 base pairs in length, encoding a protein of 458 amino acids, and shows high expression in tissues such as the lung, spleen, testis, and peripheral blood leukocytes. Genetic studies have highlighted its potential role in developmental disorders, such as global developmental delay and delayed speech and language development, linked to microdeletions encompassing the GTDC1 gene. Disruption of GTDC1 expression has been shown to affect neural progenitor cells and neurons, indicating its significance in neural development. Additionally, the gene holds evolutionary conservation, sharing 75% and 37% identity with its mouse and fruit fly counterparts, respectively.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: glycosyltransferase-like 1; Glycosyltransferase-like domain-containing protein 1; mannosyltransferase candidate; Mannosyltransferase-like protein Xa; Mat-Xa; QTMAN; tRNA-queuosine alpha-mannosyltransferase; unnamed protein product

          View more View less

          Gene Aliases: GTDC1; Hmat-Xa; mat-Xa; QTMAN

          View more View less

          UniProt ID: (Human) Q4AE62

          View more View less

          Entrez Gene ID: (Pig) 100152895, (Human) 79712

          View more View less

          Function(s)
          tRNA-queuosine(34) beta-mannosyltransferase activity transferase activity transferase activity, transferring glycosyl groups glycosyltransferase
          Process(es)
          tRNA modification regulation of translation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline