Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PDGFRA (CD140a) Antibodies

          Invitrogen

          CD140a (PDGFRA) Monoclonal Antibody (APA5), Super Bright™ 702, eBioscience™

          Advanced Verification
          39 Published Figures
          35 References
          View all (83) PDGFRA (CD140a) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD140a (PDGFRA) Monoclonal Antibody (APA5), Super Bright™ 702, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (39)
          • Advanced Verification (1)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 41

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD140a (PDGFRA) Antibody (67-1401-82) in Flow

          Staining of NIH/3T3 cells with 0.5 µg of Rat IgG2a K Isotype Control, Super Bright 702 (Product # 67-4321-82) (blue histogram) or 0.5 µg of CD140a (PDGF Receptor a) Monoclonal Antibody, Super Bright 702 (purple histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          67-1401-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          1.0 µg/test
          View 25 publications 25 publications
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Super Bright™ 702, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          APA5

          Conjugate

          Super Bright™ 702 Super Bright™ 702 Super Bright™ 702
          • Unconjugated
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant UV 661
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          413/702 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2717165

          Product Specific Information

          Description: The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the alpha chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues.

          Applications Reported: This APA5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This APA5 antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

          Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser

          Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

          Target Information

          PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; CD140 antigen-like family member A; MGC74795; OTTHUMP00000218656; PDGF A-chain; PDGF alpha chain; PDGF Receptor alpha; PDGF subunit A; PDGF-1; PDGF-R-alpha; pdgfr-a; PDGFR-alpha; unnamed protein product

          View more View less

          Gene Aliases: CD140a; Pdgfr-2

          View more View less

          UniProt ID: (Mouse) P26618

          View more View less

          Entrez Gene ID: (Mouse) 18595

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity platelet-derived growth factor alpha-receptor activity vascular endothelial growth factor-activated receptor activity platelet-derived growth factor receptor binding ATP binding vascular endothelial growth factor binding protein homodimerization activity phosphatidylinositol 3-kinase binding macromolecular complex binding platelet-derived growth factor binding phospholipase C activator activity
          Process(es)
          luteinization in utero embryonic development hematopoietic progenitor cell differentiation response to stress transmembrane receptor protein tyrosine kinase signaling pathway estrogen metabolic process positive regulation of cell proliferation male gonad development female gonad development anatomical structure morphogenesis organ morphogenesis negative regulation of platelet activation cell migration peptidyl-tyrosine phosphorylation signal transduction involved in regulation of gene expression extracellular matrix organization lung development adrenal gland development positive regulation of cell migration male genitalia development Leydig cell differentiation cellular response to reactive oxygen species platelet-derived growth factor receptor-alpha signaling pathway positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway wound healing odontogenesis of dentin-containing tooth protein autophosphorylation platelet-derived growth factor receptor signaling pathway positive regulation of fibroblast proliferation embryonic digestive tract morphogenesis embryonic cranial skeleton morphogenesis skeletal system morphogenesis positive regulation of peptidyl-tyrosine phosphorylation positive regulation of calcium-mediated signaling white fat cell differentiation positive regulation of chemotaxis cardiac myofibril assembly palate development face morphogenesis cell chemotaxis lung growth positive regulation of branching involved in lung morphogenesis retina vasculature development in camera-type eye positive regulation of ERK1 and ERK2 cascade platelet aggregation cellular response to amino acid stimulus metanephric glomerular capillary formation regulation of mesenchymal stem cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch