Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MERAAPSRRV PLQLLLLGGL ALLAAGVDAD VLLEACCADG HRMATHQKDC SLPYATESKE CRMVQEQCCH SQLEELHCAT GISLANEQDR CATPHGDNAS LEATFVKRCC HCCLLGRAAQ AQGQSCEYSL MVGYQCGQVF RACCVKSQET GDLDVGGLQE TDKIIEVEEE QEDPYLNDRC RGGGPCKQQC RDTGDEVVCS CFVGYQLLSD GVSCEDVNEC ITGSHSCRLG ESCINTVGSF RCQRDSSCGT GYELTEDNSC KDIDECESGI HNCLPDFICQ NTLGSFRCRP KLQCKSGFIQ
Fibulin-1 is a modular glycoprotein component of the elastic extracellular matrix fibers, basement membranes and blood. Fibulin-1 self associates as well as binds to calcium, fibronectin, laminin, nidogen and fibrinogen. These interactions, individually or in combination, may account for the observed association of Fibulin-1 with basement membranes, connective tissue, elastic fibers and fibrin clots. Fibulin-1 expression is stimulated by estrogen in ovarian cancer cell lines and has been suggested as both an agent of metastasis in ovarian cancer cells and an indicator for predicting cancer risk or aggressiveness in ovarian carcinomas. Other studies point to the inhibition of cancer cell motility with increasing exposure to Fibulin-1. The exact function of Fibulin-1 in the cell is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FIBL-1; Fibulin-1; Fibulin1; unnamed protein product
Gene Aliases: FBLN; FBLN1; FIBL1; PP213
UniProt ID: (Human) P23142
Entrez Gene ID: (Human) 2192
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support