Phospho-ERK1/ERK2 (Thr185, Tyr187) Recombinant Rabbit Monoclonal Antibody (15H10L7) - FAQs
Phospho-ERK1/ERK2 (Thr185, Tyr187) Recombinant Rabbit Monoclonal Antibody (15H10L7) - FAQs
View additional product information for Phospho-ERK1/ERK2 (Thr185, Tyr187) Recombinant Rabbit Monoclonal Antibody (15H10L7) - FAQs (700012)
1 product FAQs found
Do you offer antibodies for ERK kinases?
Yes, we have many antibodies against ERK 1 and 2 (e.g. 44-654G, 136200) and their phosphorylated forms. Examples of the latter include Cat. Nos. 44680G, 700012, 368800, and PA14607, among many others .
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.