CaptureSelect™ Biotin Anti-AAVX Conjugate, 500 μg - FAQs
CaptureSelect™ Biotin Anti-AAVX Conjugate, 500 μg - FAQs
View additional product information for CaptureSelect™ Biotin Anti-AAVX Conjugate - FAQs (7103522500, 7103522100)
1 product FAQs found
How many biotin molecules are there per antibody molecule in your CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522500)?
All our CaptureSelect biotin conjugates are directionally conjugated with a maximal degree of labeling of 2 biotin molecules per VHH.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.