Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NMDAR2B Antibodies

          Invitrogen

          NMDAR2B Polyclonal Antibody

          19 Published Figures
          31 References
          View all (63) NMDAR2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NMDAR2B Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (19)
          NMDAR2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NMDAR2B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NMDAR2B Antibody (71-8600) in ICC/IF

          Immunofluorescence analysis of NMDA RECEPTOR 2B was performed using 70% confluent log phase RSC96 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with NMDAR2B Rabbit Polyclonal Antibody (Product # 71-8600) at 2µg/mL in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NMDAR2B Antibody in Immunocytochemistry (ICC/IF)
          NMDAR2B Antibody in Immunocytochemistry (ICC/IF)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Immunocytochemistry (ICC/IF)
          NMDAR2B Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          NMDAR2B Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          NMDAR2B Antibody in Immunocytochemistry (ICC/IF)
          NMDAR2B Antibody in Immunohistochemistry (IHC)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)
          NMDAR2B Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NMDAR2B Polyclonal Antibody

          Product Details

          71-8600

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 18 publications 18 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Rabbit, Rat

          Published species

          Dog, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Fusion protein including a 251 amino acid sequence from the N-terminal portion of the rat NMDAR 2B protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2534001

          Target Information

          NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer's Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: glutamate [NMDA] receptor subunit epsilon-2; glutamate receptor; glutamate receptor, ionotropic, N-methyl D-aspartate 2B; glutamate receptor, ionotropic, NMDA2B; GRIN 2B; HGNC:4586; hNR 3; MGC142178; MGC142180; N-methyl D-aspartate receptor subtype 2B; NMDAR2B; NMDE2; NME2; NR2B

          View more View less

          Gene Aliases: GluN2B

          View more View less

          UniProt ID: (Rat) Q00960

          View more View less

          Entrez Gene ID: (Rabbit) 100353266, (Rat) 24410

          View more View less

          Function(s)
          NMDA glutamate receptor activity receptor binding interleukin-1 receptor binding ion channel activity extracellular-glutamate-gated ion channel activity cation channel activity calcium channel activity protein binding beta-catenin binding zinc ion binding ligand-gated ion channel activity glycine binding glutamate binding protein kinase binding glutamate-gated calcium ion channel activity D2 dopamine receptor binding glutamate receptor binding ionotropic glutamate receptor binding small molecule binding signaling receptor activity macromolecular complex binding cell adhesion molecule binding scaffold protein binding ligand-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential protein serine/threonine kinase binding heterocyclic compound binding transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential protein tyrosine kinase binding transmembrane signal receptor
          Process(es)
          action potential behavioral fear response response to hypoxia in utero embryonic development startle response suckling behavior response to amphetamine ion transport cation transport calcium ion transport cellular calcium ion homeostasis chemical synaptic transmission sensory organ development learning or memory learning memory long-term memory associative learning response to mechanical stimulus response to toxic substance response to carbohydrate response to manganese ion response to lead ion cellular response to magnesium starvation positive regulation of glutamate secretion response to amine calcium-mediated signaling trigeminal motor nucleus development hippocampus development cerebral cortex development auditory behavior regulation of ARF protein signal transduction response to magnesium ion multicellular organismal response to stress response to cytokine response to nicotine ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic response to cocaine fear response receptor clustering regulation of MAPK cascade response to ethanol response to antibiotic sensitization regulation of synaptic plasticity regulation of neuronal synaptic plasticity regulation of long-term neuronal synaptic plasticity behavioral response to pain detection of mechanical stimulus involved in sensory perception of pain protein heterotetramerization response to calcium ion response to methylmercury response to electrical stimulus response to other organism positive regulation of synaptic transmission, glutamatergic regulation of postsynaptic membrane potential excitatory postsynaptic potential long-term synaptic potentiation response to growth hormone response to fungicide cellular response to amino acid stimulus cellular response to manganese ion cellular response to dsRNA cellular response to growth factor stimulus cellular response to corticosterone stimulus cellular response to lipid positive regulation of inhibitory postsynaptic potential calcium ion transmembrane import into cytosol cation transmembrane transport regulation of presynaptic membrane potential regulation of cAMP/PKA signal transduction regulation of long term synaptic depression negative regulation of dendritic spine maintenance response to glycoside regulation of cation transmembrane transport cellular response to forskolin cellular response to curcumin response to hydrogen sulfide positive regulation of excitatory postsynaptic potential
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch