MicroCL 17 Microcentrifuge, ventilated, 120V, with hematocrit rotor - FAQs
MicroCL 17 Microcentrifuge, ventilated, 120V, with hematocrit rotor - FAQs
View additional product information for MicroCL 17 Microcentrifuge - FAQs (75002496)
1 product FAQs found
What rotor does the MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002496) come with?
The MicroCL 17 Microcentrifuge, Ventilated (Cat. No. 75002496) comes with a Hematocrit rotor.