PrioCHECK™ Porcine Toxoplasma Ab Kit - FAQs
PrioCHECK™ Porcine Toxoplasma Ab Kit - FAQs
View additional product information for PrioCHECK™ Porcine Toxoplasma Ab Kit - FAQs (7610230)
1 product FAQs found
Are negative PP values (see INSTRUCTIONS FOR USE here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0013858_7610230_UG_en.pdf) acceptable for the PrioCHECK Porcine Toxoplasma Ab Kit (Cat. No. 7610230)?
Yes, negative PP values are normal for this kit. In the QC panel for this kit's release, there were serum samples that gave negative % PP results.
Find additional tips, troubleshooting help, and resources within our Animal Health Support Center.