Biodyne™ B Nylon Membrane, 0.45 μm, 8 cm x 12 cm - FAQs
Biodyne™ B Nylon Membrane, 0.45 μm, 8 cm x 12 cm - FAQs
View additional product information for Biodyne™ B Nylon Membrane, 0.45 μm, 8 cm x 12 cm - FAQs (77016)
2 product FAQs found
Is your Biodyne B Nylon Membrane supplied sterile?
No, our Biodyne B Nylon Membrane is not supplied sterile.
What is the pore size of your Biodyne B Nylon Membrane?
The pore size of our Biodyne B Nylon Membrane is 0.45 µm.