FWS 제품은 특정 승인이 필요하므로 배송이 약 2-3주 더 소요됩니다.
Product Image
Thermo Scientific™

Imject™ mcKLH (in MES Buffer)

Thermo Scientific Imject Mariculture KLH is a purified keyhole limpet hemocyanin carrier protein that enables simple preparation of highly effective자세히 알아보기
Have Questions?
카탈로그 번호수량
776532 mg
카탈로그 번호 77653
제품 가격(KRW)
93,000
온라인 행사
Ends: 31-Dec-2026
109,000
할인액 16,000 (15%)
Each
수량:
2 mg
제품 가격(KRW)
93,000
온라인 행사
Ends: 31-Dec-2026
109,000
할인액 16,000 (15%)
Each
Thermo Scientific Imject Mariculture KLH is a purified keyhole limpet hemocyanin carrier protein that enables simple preparation of highly effective immunogens with peptide antigens.

Features of Imject Mariculture KLH:

• High-yield conjugation—each molecule of KLH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers
• Validated quality—purified and stabilized mariculture KLH maintains solubility in aqueous solutions, unlike traditional sources of the carrier protein
• Sustainable source—KLH is harvested from select populations of the mollusk Megathura crenulata (keyhole limpet) that are grown in mariculture, not extracted from wild populations
• Highly immunogenic—KLH has a high molecular mass (4.5 x 105 to 1.3 x 107 Daltons; aggregates of 350 and 390 kDa subunits) and elicits a stronger immune response than BSA or ovalbumin

KLH is the most popular and immunogenic carrier protein used for preparation of peptide antigens for immunization and antibody production. KLH preparations in phosphate buffer are ready for hapten-carrier conjugation via amine-reactive NHS-ester or glutaraldehyde crosslinking. Preparations in MES buffer are optimized for peptide-carrier conjugation via the carboxyl-reactive carbodiimide crosslinker called EDC (or EDAC).

Carrier proteins are large, complex molecules capable of stimulating an immune response upon injection. Successful production of antibodies specific to small antigens (i.e., peptides or drug compounds) requires that these haptens be covalently conjugated to a larger, more complex molecule (usually a protein) to make them immunogenic. Carrier proteins are chosen based on immunogenicity, solubility, and whether adequate conjugation with the carrier can be achieved.

Keyhole limpet hemocyanin (KLH) is widely used as a carrier protein for conjugation to haptens and other antigens to make them more immunogenic for the purpose of antibody production. Because of its large mass and complexity, KLH elicits a stronger immune response than other carrier proteins. Because the protein is derived from a mollusk, it is phylogenetically distant from mammalian species and less likely to produce antibodies that will cross-react with typical target samples in assays.

Imject Mariculture KLH (mcKLH) is KLH that has been harvested from limpets grown in mariculture rather than captured from the wild. Imject mcKLH provides the highest possible immunogenicity; it is purified and lyophilized in buffers that optimize its stability and solubility for hapten conjugation. As a large protein, KLH has hundreds of primary amines and carboxyl groups that can be targeted for conjugation with NHS-ester, EDC, and other crosslinking reagents.

The carbodiimide crosslinker EDC conjugates carboxyl-containing haptens (e.g., C-terminus of peptide antigens) to the highly immunogenic KLH carrier protein for immunization and antibody production. This method of immunogen preparation is ideal for peptide antigens with few or no aspartic and glutamic acid residues (carboxylates) and lysine residues (primary amines) within the central portion of the primary sequence. Because peptides contain both carboxylate and amines, EDC conjugation results in their becoming variously polymerized and randomly oriented in their linkage to the carrier protein. Typically, this results in a high level of antigen loading on the carrier protein as well as presentation in all possible orientations for antibody production. However, important (desired) epitopes within the antigen peptide sequence may be blocked by EDC-mediated conjugation if those regions contain primary amines (lysine residues) or carboxylates (aspartic and glutamic acid residues). In these cases, either use a homobifunctional amine-reactive crosslinker with the purified mcKLH in phosphate buffer or synthesize the peptide with a unique terminal cysteine and use a kit with Malemide-Activated mcKLH to prepare the carrier protein conjugate.

More Product Data
• Carrier protein activation and conjugation data for immunogen preparation

Related Products
Imject™ mcKLH (in PBS)
Imject™ mcKLH Subunits, High Purity Research Grade
Imject™ EDC mcKLH Spin Kit
Imject™ Purification Buffer Salts
For Research Use Only. Not for use in diagnostic procedures.
사양
설명Imject mcKLH (in MES buffer)
수량2 mg
제품라인Imject
Unit SizeEach
구성 및 보관
Upon receipt store product at 4°C. Product is shipped at ambient temperature.

자주 묻는 질문(FAQ)

What do I need to consider when designing a peptide for antibody production?

We recommend that hydrophobic residues comprise 50% or less of all the residues in your sequence. Make sure there is at least one charged residue for every five amino acids: this is generally known to enhance the solubility of the peptide. Peptides (compared to polypeptides, which fold and bury the hydrophobic amino acids) are too small to fold, so just a few hydrophobic amino acids may leave them insoluble.

Other guidelines include:
Peptides containing multiple Cys, Met, and Trp can be hard to synthesize.
Some sequences are problematic in solid-phase peptide synthesis or cleavage and are best avoided; these include Asp-Pro sequences or stretches of amino acids that require bulky protecting groups on their side chains during synthesis.
Glycine is often good for antigenicity as it has only a hydrogen side chain; this allows for complete rotation.
Please note that we offer a Custom Antibody Production service (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production.html) that includes the use of our proprietary Antigen Profiler and Antigen Preparation tool (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production/antigen-profiler-antigen-preparation.html).

What is the optimal peptide length for antibody production?

Most peptide antigens range in length from 12 to 16 residues and are relatively easy to synthesize. Peptides of 9 residues or shorter have been effective antigens for antibody production, but peptides longer than 16 amino acids may contain several epitopes and form secondary structures. Peptides in excess of 18 residues begin to present more synthetic challenges. Before you synthesize your peptide, we recommend doing a BLASTP search using your peptide sequence. This is to make sure that the peptide is not homologous or identical to a sequence in a completely unrelated protein in the host animal.

Please note that we offer a Custom Antibody Production service (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production.html) that includes the use of our proprietary Antigen Profiler and Antigen Preparation tool (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production/antigen-profiler-antigen-preparation.html).

What happens if antibodies are stored improperly?

Improper storage of antibodies can lead to:
- Degradation: Loss of activity and specificity
- Aggregation: Formation of precipitates or clumps
- Reduced performance: Poor results in assays or experiments

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

What do antibody pair kits contain, how many samples can I process, and how can I find a list of antibody pair kits by target?

Antibody pair kits contain capture antibody, detection antibody, recombinant standard and HRP conjugate. Each contains enough reagents to process forty 96-well plates. A list of Antibody Pair Kits (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/elisa/antibody-pair-kits.html) is available by target.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

What do Reagent Sets contain, how many samples can I process, and how can I check if a Reagent Set is available for my target?

Reagent Sets contain Capture Antibody, Detection Antibody, Recombinant Standard, HRP Conjugate, TMB Substrate and Stop Solution. Each contains enough reagents to process five 96-well plates. Reagent Sets are included in the main list of ELISA (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/elisa/antibody-pair-kits.html) kits (search by “Reagent Set”).

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.