Which protein assays are the T-PER Tissue Protein Extraction Reagent (Cat. No. 78510) compatible with?
The T-PER Tissue Protein Extraction Reagent (Cat. No. 78510) is compatible with standard protein assays such as Pierce BCA Protein Assay (Cat. No. 23225) and Coomassie Plus (Bradford) Assay Kit (Cat. No. 23236).