Search
Search
7TM Antibodies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Vasopressin receptor 1a (AVPR1A) is a receptor for arginine vasopressin. The activity of this receptor is mediated by G-proteins which activate a phosphatidyl-inositol-calcium second messenger system. AVPR1A has been involved in social behaviors, including affiliation and attachment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antidiuretic hormone receptor 1a; AVPR V1a; G-protein coupled receptor; AVPR1A; SCCL vasopressin subtype 1a receptor; V1-vascular vasopressin receptor AVPR1A; V1a receptor; V1a vasopressin receptor; V1aR; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor; Vasopressin V1aR
Gene Aliases: AVPR V1a; AVPR1; AVPR1A; V1aR
UniProt ID: (Human) P37288
Entrez Gene ID: (Human) 552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support