CaptureSelect™ tPA Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ tPA Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ tPA Ligand Leakage ELISA is designed to detect possible leached ligand of tissue plasminogen activator (tPA) purified using CaptureSelect tPA Affinity Matrix.
製品番号(カタログ番号)数量
8103430011 Assay
81034301010 Assays
製品番号(カタログ番号) 810343001
価格(JPY)
171,300
Each
数量:
1 Assay

CaptureSelect tPA Ligand Leakage ELISA is designed to detect possible leached ligand of tissue plasminogen activator (tPA) purified using CaptureSelect tPA Affinity Matrix. The ligand, a 13 kD recombinant protein, is covalently coupled to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect tPA Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
仕様
検出限界1 ng/mL
フォーマットIndividual components
製品ラインCaptureSelect
製品タイプLigand Leakage ELISA
数量1 Assay
出荷条件Dry Ice
ターゲットCaptureSelect tPA Affinity Ligand
検出法Absorbance at 450 nm
イムノアッセイキットのフォーマットSandwich ELISA Kit
サンプルタイプLigand Leachate
ターゲット種Not Applicable
Unit SizeEach
組成および保存条件
Store at -20°C.