L-Proline for SILAC
Thermo Scientific™

L-Proline for SILAC

Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
88211115 mg
88430500 mg
Catalog number 88211
Price (USD)
59.25
Each
Quantity:
115 mg
Request bulk or custom format
Price (USD)
59.25
Each

Thermo Scientific Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits. This is a lyophilized preparation of L-Proline for SILAC experiments.

General features of heavy and light amino acids for SILAC labeling:

  • Efficient—100% label incorporation into proteins of living cells
  • Flexible—different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
  • Multiplex capabilities—several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
  • High-quality supplements—heavy amino acids with >99% isotope purity

Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Thermo Scientific Heavy and Light Amino Acids for SILAC are used together with specialized cell culture media that are deficient in essential amino acids. Heavy and light L-lysine and L-arginine are the most common amino acids used for SILAC analysis of tryptic peptides. Up to three different experimental conditions can be readily analyzed with different isotopes of lysine and arginine. For lysine three-plex experiments, 4,4,5,5-D4 L-lysine and 13C6 15N2 L-lysine are used to generate peptides with 4- and 8-Da mass shifts, respectively, compared to peptides generated with light lysine. For arginine three-plex experiments, 13C6 L-arginine and 13C6 15N4 L-arginine are used to generate peptides with 6- and 10-Da mass shifts, respectively, compared to peptides generated with light arginine. L-leucine is another amino acid commonly used for SILAC labeling, because it is one of the most common amino acids found in protein sequences. Proline is a non-essential amino acid that is sometimes added to SILAC media to prevent the metabolic conversion of heavy arginine to heavy proline in mammalian cell lines with high arginine dehydrogenase activity.

Related Products

  • L-Arginine-HCl for SILAC (Cat. No. 89989)
  • L-Leucine for SILAC (Cat. No. 88428)
  • L-Lysine-2HCl for SILAC (Cat. No. 89987)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeMass Isotope of Amino Acid
Quantity115 mg
Shipping ConditionRoom Temperature
Workflow StepProtein Labeling
Detection MethodMass Spectrometry
FormPowder
Product TypeAmino Acids
Starting MaterialCells
Unit SizeEach
Contents & Storage
Store at room temperature.