Pierce™ Glycoprotein Isolation Kit, WGA - FAQs
Pierce™ Glycoprotein Isolation Kit, WGA - FAQs
View additional product information for Pierce™ Glycoprotein Isolation Kit, WGA - FAQs (89805)
2 product FAQs found
What is the difference between your Pierce Glycoprotein Isolation Kit, ConA and WGA?
The ConA kit (Cat. No. 89804), which has concanavalin A immobilized on the resin, recognizes alpha-linked mannose, and to a lesser extent, terminal glucose residues. The WGA kit (Cat. No. 89805), which has wheat germ agglutinin immobilized on the resin, recognizes N-acetyl glucosamine (GlcNAc) groups and sialic acid.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I isolate my proteins based on their post-translational modification (PTM) or active binding site?
Enrichment of specific proteins or protein complexes can most easily be accomplished by using target-specific immobilized antibody or PTM specific affinity binding. We offer several kits targeting phosphorylated proteins, alpha- linked mannose and terminal glucose residues, N-acetyl glucosamine, polyubiquinated proteins, ATP binding sites, and active serine hydrolase enzymes. Choose the right protein enrichment kit for your application using the following link (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/protein-enrichment.html).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.