See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Microplates
▸
Filtering Microplates
▸
Collection Plates (Polypropylene) for Pierce™ 96-Well Filter Spin Plates
Thermo Scientific™
Collection Plates (Polypropylene) for Pierce™ 96-Well Filter Spin Plates
Collection plates and balance plates for use with 96-well filter spin plates
Technical Support
Customer Service
Catalog Number
Quantity
89813
6 plates
Catalog number
89813
Price (GBP)
54.25
Each
Add to cart
Quantity:
6 plates
Request bulk or custom format
Price (GBP)
54.25
Each
Add to cart
These Thermo Scientific Plate Accessories are collection plates and centrifuge balance plates for affinity purification and desalting resin products that use Thermo Scientific Pierce 96-Well Filter Spin Plates.
Related Products
Balance Plate for Pierce™ 96-Well Filter Spin Plates
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Plate Compatibility
96-Well Plates
Product Type
Collection Plate
Quantity
6 plates
Material
Polypropylene
No. of Wells
96
Product Line
Pierce
Unit Size
Each
Contents & Storage
0.0