Balance Plate for Pierce™ 96-Well Filter Spin Plates
Balance Plate for Pierce™ 96-Well Filter Spin Plates
Balance Plate for Pierce™ 96-Well Filter Spin Plates
Thermo Scientific™
Balance Plate for Pierce™ 96-Well Filter Spin Plates
These Thermo Scientific Plate Accessories are collection plates and centrifuge balance plates for affinity purification and desalting resin products thatRead more
Have Questions?
Catalog NumberQuantity
452051 plate
Catalog number 45205
Price (TWD)
497.00
Online offer
Ends: 30-Sep-2026
710.00
Save 213.00 (30%)
Each
Quantity:
1 plate
Price (TWD)
497.00
Online offer
Ends: 30-Sep-2026
710.00
Save 213.00 (30%)
Each
These Thermo Scientific Plate Accessories are collection plates and centrifuge balance plates for affinity purification and desalting resin products that use Thermo Scientific Pierce 96-Well Filter Spin Plates.
Related Products
Collection Plates (Polypropylene) for Pierce™ 96-Well Filter Spin Plates
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity96-Well Plate
DisposableSingle-use
Product LinePierce
Quantity1 plate
For Use With (Equipment)96-well Plate
Product TypeBalance Plate
Unit SizeEach