What are the contents of the GeneChip Hybridization Control Kits?
We offer two GeneChip Hybridization Control Kits, that differ in size:
- Cat. No. 900454: 30 rxns
- Cat. No. 900457: 150 rxns
Both contain 20X Hybridization Control Shock and Control Oligo B2.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
- Cat. No. 900454: 30 rxns
- Cat. No. 900457: 150 rxns
Both contain 20X Hybridization Control Shock and Control Oligo B2.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.