Product Image
Applied Biosystems™
B2 Control Oligo (3nM)
The B2 Control Oligo (3nM) is part of the GeneChip™ Hybridization Control Kit, but may be purchased separately.
Have Questions?
Número de catálogoCantidad
9003011 each
Número de catálogo 900301
Precio (MXN)
-
Cantidad:
1 each
The B2 Control Oligo (3nM) is part of the GeneChip™ Hybridization Control Kit, but may be purchased separately.
The Hybridization Control Kit is required for use with GeneChip brand eukaryotic expression arrays.
Related Links:
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Clasificación de la sondaEukaryotic Expression Probe
Tipo de productoOligonucleótidos, cebadores
Cantidad1 each
Unit SizeEach
Preguntas frecuentes
What are the contents of the GeneChip Hybridization Control Kits?
We offer two GeneChip Hybridization Control Kits, that differ in size:
- Cat. No. 900454: 30 rxns
- Cat. No. 900457: 150 rxns
Both contain 20X Hybridization Control Shock and Control Oligo B2.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What signal and fold-change concordance can I expect between Affymetrix miRNA 4.0 and miRNA 4.1 Arrays and miRNA 3.0 and miRNA 3.1 Arrays?
You should expect high signal and fold-change concordance for transcripts with identical probes on the miRNA 4.0 and miRNA 4.1 Arrays and the miRNA 3.0 and miRNA 3.1 Arrays. The snoRNA, scaRNA, and precursor miRNA probe sets were reselected, and for some probe sets in these categories, the probes are not identical between the two array designs. Because the probes selected may be different, one may observe changes in the probe set signal summary for a given target. This is possible as different probe sequences will have varying affinity to the target and may show a higher or lower absolute probe signal depending on the GC content of the probes. However, fold changes are typically well-correlated to the legacy probe set unless there is cross-hybridization in the legacy probe set. Our expectation is that more unique and specific probes result in more specific hybridization to the intended target, and therefore a better performing probe set. Target preparation replicates of human brain and lung samples were pooled and hybridized to four miRNA 3.0 Arrays and four miRNA 4.0 Arrays. The Pearson product moment correlation was calculated from the median signal of matched probe sets and from the median fold change of detected matched probe sets on the miRNA 3.0 Array compared to the miRNA 4.0 Array. Probe sets were defined as detected if the median DABG p-value of the four miRNA 3.0 Array brain replicates was less than 0.06. Two subsets of matched probe sets were used for miRNA 3.0 Array to miRNA 4.0 Array signal and fold change correlation: All human probe sets including those that do not necessarily share identical probe sequences and human probe sets for which the probe sequences are identical on both the miRNA 3.0 and miRNA 4.0 Arrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Do we have to use any Affymetrix Reagents with our assay?
GeneChip Hybridization Control Kit 30 reactions
GeneTitan Hybridization, Wash, and Stain Kit for miRNA Array Plates 96 reactions
GeneTitan Hybridization Module for miRNA Plates 96 reactions
Affymetrix FlashTag Biotin HSR RNA Labeling Kit 10 reactions
Affymetrix FlashTag Biotin HSR RNA Labeling Kit 30 reactions
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What kind of dynamic range can I expect from Affymetrix miRNA Array Plates?
Affymetrix miRNA Array Plate provides a similar dynamic range to GeneChip miRNA Array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do results from GeneChip miRNA Cartridge Array compare to Affymetrix miRNA Array Plates?
When evaluating non-control probe sets in common to both arrays, we observed very high signal and fold-change correlation for GeneChip miRNA Cartridge Arrays compared to Affymetrix miRNA Array Plates (Pearson correlation coefficients of 0.97-0.98 for signal correlation and Pearson correlation coefficients of 0.96-0.98 for fold-change correlation).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.