Product Image
Applied Biosystems™
GeneChip™ DNA Labeling Reagent, 7.5 mM
The GeneChip™ DNA Labeling Reagent is used with the Prokaryotic Target Labeling Assay for 3' expression studies with GeneChip prokaryoticRead more
Have Questions?
Catalog NumberQuantity
9005421 kit
Catalog number 900542
Price (USD)
536.00
Each
Quantity:
1 kit
Price (USD)
536.00
Each
The GeneChip™ DNA Labeling Reagent is used with the Prokaryotic Target Labeling Assay for 3' expression studies with GeneChip prokaryotic Brand Arrays. The assay utilizes reverse transcriptase and random hexamer primers to produce DNA complementary to the RNA. The cDNA products are then fragmented by DNase I and labeled with terminal transferase and biotinylated GeneChip DNA Labeling Reagent at the 3' termini.
Functionally tested on GeneChip Brand Arrays, we recommend the Prokaryotic Target Labeling Assay and associated reagents for use in prokaryotic gene expression studies.
Prokaryotic Target Labeling Assay components include:
• Control Oligo B2, 3nM
• GeneChip DNA Labeling Reagent
• GeneChip Poly-A RNA Control Kit
Detailed instructions for using this reagent are described in Section 3, Chapter 1 of the GeneChip Expression Analysis Technical Manual.
Each kit contains 1 vial of 60 μL of the GeneChip DNA Labeling Reagent, 7.5 mM, sufficient for 30 labeling reactions.
Related Links
GeneChip™ Wash Buffer A
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes Label or DyeDirect Labeling
Product LineGeneChip
Product TypeDNA Labeling Reagent
Quantity1 kit
Detection MethodBiotin-based
Final Product TypecDNA (Labeled)
Labeling TargetRNA (General)
Unit SizeEach
Frequently asked questions (FAQs)
What is the shelf life of this the DNA Labeling reagent (Cat. No. 900542)?
The DNA Labeling Reagent has about 12 months of dating from the date of shipment.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.