Product Image
Applied Biosystems™

GeneChip™ Eukaryotic Poly-A RNA Control Kit

The GeneChip™ Eukaryotic Poly-A RNA Control Kit is used with the Prokaryotic Target Labeling Assay and the Prokaryotic Target Labeling Assay for 3' expression studies with prokaryotic GeneChip Brand Arrays.
Have Questions?
Número de catálogoCantidad
9004331 kit
Número de catálogo 900433
Precio (MXN)
-
Cantidad:
1 kit

The GeneChip™ Eukaryotic Poly-A RNA Control Kit is used with the Prokaryotic Target Labeling Assay and the Prokaryotic Target Labeling Assay for 3' expression studies with prokaryotic GeneChip Brand Arrays. The assays utilize reverse transcriptase and random hexamer primers to produce DNA complementary to the RNA. The cDNA products are then fragmented by DNase I and labeled with terminal transferase and biotinylated GeneChip DNA Labeling Reagent at the 3' termini.

Functionally tested on GeneChip Brand Arrays, we recommend the Prokaryotic Target Labeling Assay and associated reagents for use in prokaryotic gene expression studies.

Prokaryotic Target Labeling Assay components include:

  • B2 Control Oligo (3nM)
  • GeneChip DNA Labeling Reagent
  • GeneChip Poly-A RNA Control Kit

Related Links:

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Tipo de productoOligonucleótidos, cebadores
Cantidad1 kit
PrimerRandom Hexamer
Línea de productosGeneChip
Unit SizeEach

Preguntas frecuentes

What are the estimated Poly-A RNA Control final concentrations when processing expression arrays?

Here are the Poly-A RNA Controls final concentrations (in samples):
- lys 1:100,000
- phe 1:50,000
- thr 1:25,000
- dap 1:7,500

What is the shelf life of the Poly-A RNA Control's first dilution during processing of expression microarrays?

The first dilution of the Poly-A RNA Control can be stored at -20 degrees C for 6 weeks (with up to 4 freeze/thaws).

Can I purchase one, or all, of the four Poly-A Controls separately?

These four controls are only available as a pre-mixed cocktail with the defined concentrations and relative ratios. This combination of concentrations has been tested to provide qualitative information on both sensitivity and dynamic range of the labeling assay. Providing these controls in a pre-mixed format increases ease-of-use and reduces the number of manual dilution steps.
Information on ordering individual poly-A control clones and preparing poly-A stocks is available in Section 2, Chapter 2, and Section 3, Chapter 2, of the GeneChip Expression Analysis Technical Manual.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Can I purchase the Poly-A Control Stock and the Poly-A Control Dil Buffer separately?

The Control Stock and the Dilution Buffer are packaged in the same Kit to provide convenience to customers. Currently they cannot be purchased as separate products.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Do you have any specific recommendation on the type of non-stick tubes to use for diluting the Poly-A RNA Control Stock?

The following tubes were used in-house during product development of this Poly-A RNA Control Kit, and we have not tested any other tubes at this time:
Non-stick RNase-free 0.5 mL microfuge tubes, Ambion, Cat. No. 12350
Non-stick RNase-free 1.5 mL microfuge tubes, Ambion, Cat. No. 12450

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.