Axiom™ Genome-Wide BOS 1 Bovine Array - FAQs
Axiom™ Genome-Wide BOS 1 Bovine Array - FAQs
View additional product information for Axiom™ Genome-Wide BOS 1 Bovine Array - FAQs (901791)
18 product FAQs found
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are there any service providers for the Axiom Genome-Wide BOS 1 Array Plate?
A number of service providers have experience running the Axiom Assay and the genotyping call algorithm that is used with the Axiom Genome-Wide BOS 1 Array Plate.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What size are the files associated with the Axiom Genome-Wide BOS 1 Array Plate?
The average sample file size is 28 MB.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What quality control tools are available for the Axiom Genome-Wide BOS 1 Array?
Dish QC (DQC) is the recommended QC metric for Axiom Genome-Wide Arrays in Genotyping Console Software. The default threshold is greater than or equal to 0.82 for each sample. It operates by measuring signal at a collection of sites in the genome that are known not to vary from one individual to the next. Because the metric monitors non-polymorphic locations, at each position it is known which of the two channels in the assay should contain signal and which should be just background. DQC is a measure of the extent to which the distribution of signal values is separated from background values, with 0 indicating no separation and 1 indicating perfect separation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I analyze the data generated by the Axiom Genome-Wide BOS 1 Array Plate?
Axiom Analysis Suite is used to analyze data from the Axiom Genome-Wide BOS 1 Array Plate.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What types of genomic DNA were tested with the Axiom Genome-Wide BOS 1 Array and assay?
A variety of samples, including those originating from blood, semen, nasal swabs, hair bulbs, and ear punch tissue, were tested using the Axiom Genome-Wide BOS 1 Array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How much and what quality of genomic DNA is required to perform the Axiom Genome-Wide BOS 1 assay?
The assay requires 200 ng of high-quality, double-stranded genomic DNA that is not highly degraded. Genomic DNA must be of high purity and free of DNA polymerase inhibitors such as high salt, hemes, and chelating agents. For details about general assay requirements for genomic DNA, please refer to Chapter 2 of the Axiom 2.0 Genotyping Assay User Guide (https://tools.thermofisher.com/content/sfs/manuals/axiom_genotyping_solution_analysis_guide.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What assay kit do I use with the Axiom Genome-Wide BOS 1 Array Plate?
The Axiom Genome-Wide BOS 1 Array Plate is designed for use with the Axiom 2.0 Reagent Kit. Assay kits are available for 96 (one plate), 192 (two plates), and 768 (eight plates) reactions.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What performance can I expect from samples obtained from different cattle breeds for the Axiom Genome-Wide BOS 1 Array?
The performance of the array was measured across 20 breeds, and the average sample call rate is 99.62 percent.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the reproducibility of SNP calls for replicates run in different plates using the Axiom Genotyping Assay?
The reproducibility for the validated SNPs on the screening arrays is 99.94 percent.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How reliable are the SNP calls for the Axiom Genome-Wide BOS 1 Array?
The Mendelian consistency rate of the validated SNPs on the Axiom Genome-Wide BOS 1 Array Plate is 99.96 percent among different experiments.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can the Axiom Genome-Wide BOS 1 Array Plate be used for copy number analysis?
There are no copy number probes on the array. However, measuring copy number variation (CNV) in the bovine genome can be performed using a custom whole-genome sampling assay design (a cartridge-based SNP Array 6.0 genotyping assay).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How was the LD metric calculated with regard to breed for the Axiom Genome-Wide BOS 1 Array?
LD calculations were carried out for each breed separately.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How was genetic coverage calculated for the Axiom Genome-Wide BOS 1 Array?
Genetic coverage was calculated based on SNP pairwise LD (r2) results obtained from genotype data of reasonably unrelated samples within each breed of interest. The default r2 threshold for genetic coverage was 0.8. Unless otherwise specified, all SNPs with r2 results for a given breed are included in the target coverage set. This set includes all SNPs that are converted in the screen and have polymorphic genotype data for the breed of interest. For breeds for which bovine HapMap genotype data were available for enough of the samples used in the screen, HapMap SNPs with polymorphic genotypes were also included in the target coverage set.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How were SNPs selected for the Axiom Genome-Wide BOS 1 Array?
SNPs were selected to represent polymorphisms from a comprehensive set of commercially important breeds of dairy and beef cattle from both Bos indicus and Bos taurus. Our primary goal was to obtain high levels of genetic coverage. SNP pairwise linkage disequilibrium (LD) values (r2) were calculated to identify SNPs in strong LD. Then we selected the fewest number of SNPs to cover the known genetic variation for each breed we prioritized (first five breeds in Table 1). After SNPs were selected for optimal genetic coverage, the distances between the selected SNPs were calculated, and inter-SNP gaps filled (starting with the largest) by selecting polymorphic SNPs from the five major breeds: Holstein, Angus, Nelore, Jersey, and Fleckvieh.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What were the steps in designing the Axiom Genome-Wide BOS 1 Array?
We obtained information for more than 46 million SNPs from sequencing efforts coordinated by the Affymetrix Bovine Consortium (basic and applied researchers in the bovine community). From these, we selected SNPs to use based on physical coverage of the bovine genome and the number of breeds in which the SNPs were observed.
We then screened many millions of SNPs against approximately 400 samples from the Affymetrix Bovine Consortium and bovine HapMap samples (Texas A&M University). The result is a database of ~3 million validated SNPs, which means each has been demonstrated as a real, truly polymorphic SNP and has been proven to work in the assay. This implies that customers should be able to select SNPs from our database and create their own custom array, and that the SNPs chosen should perform at a very high level in the assay.
Finally, we selected a subset of more than 648,000 SNPs primarily based on genetic coverage for a selection of breeds and secondarily based on physical coverage of the bovine genome.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How many SNPs are tiled on the Axiom Genome-Wide BOS 1 Array?
Approximately 648,000 SNPs are tiled on the array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.