Axiom™ Genome-Wide BOS 1 Bovine Array
Applied Biosystems™

Axiom™ Genome-Wide BOS 1 Bovine Array

The Axiom Genome-Wide BOS 1 Array Plate is designed to maximize genetic and physical SNP coverage and offers up toRead more
Have Questions?
Catalog NumberQuantity
9017911 plate
Catalog number 901791
Price (USD)
-
Quantity:
1 plate
The Axiom Genome-Wide BOS 1 Array Plate is designed to maximize genetic and physical SNP coverage and offers up to 34 percent more genetic coverage of commercially important cattle breeds, including Bos taurus, Bos indicus, as well as dairy and beef breeds. The array covers more than 640,000 validated SNP markers representing the genetic diversity of approximately 3 million SNPs from the Affymetrix Bovine Genomic Database.

Developed in collaboration with key scientists in the bovine genotyping industry, the Axiom Genome-Wide BOS 1 Array Plate enables accurate genetic merit evaluations, genome-wide association studies to identify variations associated with disease, drug response, and other economically important traits, as well as biodiversity research and linkage disequilibrium (LD) studies.

Benefits of the Axiom Genome-Wide BOS 1 Array Plate:
• Up to 34 percent higher genetic coverage than any array on the market
• The broadest coverage of beef and dairy breeds
• Designed and priced with breeders in mind
• Built from a database of 3 million validated SNPs for reliable results
• Developed in collaboration with 10 leading bovine researchers

Note: Validated SNPs are SNPs that are polymorphic among 20 cattle breeds and demonstrate high performance on the Axiom genotyping assay.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesBovine
Unit SizeEach

Frequently asked questions (FAQs)

Are there any service providers for the Axiom Genome-Wide BOS 1 Array Plate?

A number of service providers have experience running the Axiom Assay and the genotyping call algorithm that is used with the Axiom Genome-Wide BOS 1 Array Plate.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What size are the files associated with the Axiom Genome-Wide BOS 1 Array Plate?

The average sample file size is 28 MB.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What quality control tools are available for the Axiom Genome-Wide BOS 1 Array?

Dish QC (DQC) is the recommended QC metric for Axiom Genome-Wide Arrays in Genotyping Console Software. The default threshold is greater than or equal to 0.82 for each sample. It operates by measuring signal at a collection of sites in the genome that are known not to vary from one individual to the next. Because the metric monitors non-polymorphic locations, at each position it is known which of the two channels in the assay should contain signal and which should be just background. DQC is a measure of the extent to which the distribution of signal values is separated from background values, with 0 indicating no separation and 1 indicating perfect separation.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I analyze the data generated by the Axiom Genome-Wide BOS 1 Array Plate?

Axiom Analysis Suite is used to analyze data from the Axiom Genome-Wide BOS 1 Array Plate.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What types of genomic DNA were tested with the Axiom Genome-Wide BOS 1 Array and assay?

A variety of samples, including those originating from blood, semen, nasal swabs, hair bulbs, and ear punch tissue, were tested using the Axiom Genome-Wide BOS 1 Array.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.