CytoScan™ HD Array Kit and Reagent Kit Bundle - FAQs
CytoScan™ HD Array Kit and Reagent Kit Bundle - FAQs
View additional product information for CytoScan™ HD Array Kit and Reagent Kit Bundle - FAQs (901835)
124 product FAQs found
Which thermocycler can I use for the CytoScan HD assay?
Here is the link to the CytoScan assay user guide: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/703038_cytoscan_assay_UG.pdf
In Appendix D (starting on page 67), the manual outlines specifications for compatible thermocyclers. Any thermocycler that fits those specifications will work.
Note: Use the silver or gold-plated block only, do not use an aluminum block. In order to determine which block you have, look at the serial number on the block. An "A" in the serial number indicates that the block is aluminum; an "S" indicates that it is silver. Also, an aluminum block has a honeycomb appearance between the wells, whereas a silver block is smooth.
What are the recommended genomic DNA extraction methods for CytoScan HD assay?
Methods that include boiling or strong denaturants are not acceptable, because the DNA would be rendered single-stranded. For genomic DNA extraction, we recommend using either the Qiagen Gentra Puregene Kit or the 5 Prime PerfectPure DNA Blood Kit.
Our CytoScan HD Assay requires genomic DNA concentration ≥50 ng/µL. Therefore, the elution volumes for each of the kits will need to be adjusted accordingly to achieve the desired concentration.
The presence of RNA and free nucleotides can interfere with some quantitation methods using a spectrophotometer or a NanoDrop instrument. To eliminate RNA contamination, perform RNase treatment during extraction as follows:
- For QIAGEN Gentra Puregene Kit: Perform RNase treatment as recommended in the extraction kit manual prior to elution of genomic DNA.
- For 5 PRIME PerfectPure DNA Blood Kit: Use only RNase-treated purification columns for extraction of genomic DNA.
The purified genomic DNA extracted using the two methods above should meet the DNA quality specifications per the manufacturer’s kit extraction manual.
Note: Both blood and cell line sample sources have been tested for the CytoScan HD assay.
What are the specifications for the CytoScan HD Array?
The CytoScan HD Array includes 750,000 "genotype-able" SNPs and 1.9 million non-polymorphic probes. The CytoScan HD Array covers both constitutional and cancer genes with: - ISCA constitutional coverage at 1 marker /384 bases
- Complete cancer gene coverage at 1 marker / 553 bases
- 12,000 OMIM genes at 1 marker / 659 bases
- >36,000 RefSeq genes with 1 marker / 880 bases
- Backbone (non-gene) coverage of 1 marker / 1,737 bases across genome for breakpoints
Array format: 49
Feature size: 5 µM
Time to scan per array: ~32 mins
Hyb temperature: 50 degrees C
The CytoScan HD Array uses the following QC metrics:
- SNPQC ≥15
- MAPD ≤0.25
- Waviness SD ≤0.12
I accidentally froze the wash buffers in my CytoScan HD Kit. Can I still use them for purification?
The Purification Wash Buffer and Elution Buffer are not affected by being frozen at -20 degrees C and are safe to use for purification.
How is gender determined by the Cytoscan HD array?
A subset of Y probes is used and run through a Hidden Markov Model to determine gender. Copy number of 0 or 1. 0 indicates female and 1 indicates male.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the Minor Allele Frequency of an individual marker on CytoScan HD?
The Minor Allele Frequency (MAF) refers to the frequency at which the least common allele occurs in a given population.
For example: A SNP with an allele (G) frequency of 0.40 implies that 40% of the population has G allele versus the most common allele (major allele), which is found in 60% of the population. If an SNP allele frequency is 0.05 in a population of 100 people, the number of alleles for each SNP is: 100 * 2 = 200 (each individual has two alleles).
Total number alleles for your SNP with a MAF of 0.05: 200 *0.05 = 10.
The minor allele is present ten times in your population. However, you don't know how many are heterozygotes vs homozygotes.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why does the CytoScan Assay User Guide state on page 35 to always prepare Fragmentation Master Mix according to Table 9 even when processing less than 24 samples?
It is critical to have correct size of the fragments for an optimal hybridization. The fragmentation reagent is very viscous, and it is easier to get a correct proportion of enzyme when a larger volume is used. Therefore, our recommendation is to always prepare a master mix for 24 samples, even if a smaller number of samples are processed.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Do you offer probes in the CytoScan HD array that cover the mitochondrial genome?
No, we do not offer probes in the Cytoscan HD array that cover the mitochondrial genome.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the SNP overlap between CytoScan HD Array and Genome-Wide Human SNP Array 6.0 (Cat. No. 901182)?
There are 549,673 SNPs that are genotyped on CytoScanHD which match to a marker on the Genome-Wide Human SNP Array 6.0.
Note: CytoScan HD Array can be ordered as part of the following kits: CytoScan HD Array Kit and Reagent Kit Bundle (Cat. No. 901835) and CytoScan HD Training Kit (Cat. No. 901834)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the pre-amplification clean room requirements for Axiom assays?
When manipulating genomic DNA, precautions need to be taken to avoid contamination with foreign DNA amplified in other reactions and procedures.
- We recommend that genomic DNA manipulations be performed in a dedicated pre-amplification room or area separate from the main laboratory. This pre-amplification area should have a dedicated set of pipettes and plasticware.
- If a dedicated area is not available, we recommend using a dedicated bench or a dedicated biosafety hood with dedicated pipettes.
- If a dedicated bench or biosafety hood is not available, a set of dedicated pipettes is recommended at the least.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which thermal cyclers have been tested for use with Cytoscan HD?
We recommend using the following thermal cyclers with Cytoscan HD:
For pre-PCR, we recommend 2720 Thermal Cycler (Cat. No. 4359659)
For pre- and post-PCR , we recommend the following thermal cyclers:
- GeneAmp PCR System 9700 Gold (Cat. No. 4314878) (Note: This instrument has been discontinued and therefore cannot be ordered)
- GeneAmp PCR System 9700 Silver (Cat. No. N8050001) (Note: This instrument has been discontinued and therefore cannot be ordered)
- Applied Biosystems Veriti Dx 96-Well Thermal Cycler (Cat. No. 4452300)
- Applied Biosystems Veriti 96-Well Thermal Cycler (Cat. No. 4375786)
- Bio-Rad® DNA Engine® PTC-200 Thermal Cycler
- Eppendorf® Mastercycler® Pro S Thermal Cycler
Note: You must use thermal cyclers with silver or gold-plated silver blocks. Do not use thermal cyclers with aluminum blocks.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why is it important to use Tough-Spots® labels when using GeneChip cartridge arrays?
Tough-Spots® labels are small adhesive stickers used to temporarily seal the backs of cartridge arrays during the overnight hybridization step. They are required to prevent loss of volume due to evaporation through the septa. We recommend using Tough-Spots® labels on Rolls from USA Scientific (Item No. 9185-0000)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the respective hybridization volumes based on array format for Affymetrix gene expression arrays?
Proper hybridization volume is critical to obtaining an even signal across a given array. Too little volume can lead to black circles in the middle of the array. Too much volume can leak out of the back of the array. The correct hybridization volume leaves enough room for a small air bubble to circulate around the array surface during the overnight hybridization. Here are the recommended hybridization and fill volumes based on the array format:
Array Format; Hybridization Volume; Fill Volume
- 49 Format (Standard); 200 µL; 250 µL
- 64 Format; 200 µL; 250 µL
- 100 Format (Midi); 130 µL; 160 µL
- 169 Format (Mini); 80 µL; 100 µL
- 400 Format (Micro); 80 µL; 100µL
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the hybridization temperature and rotation speed for CytoScan arrays?
The hybridization temperature is 50 degrees C. The rotation speed is 60 rpm.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the array QC metrics for the CytoScan HD arrays?
Cytoscan HD arrays use the following QC metrics: SNPQC≥15, MAPD≤0.25, Waviness-SD≤0.12.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the recommended requirements for genomic DNA samples used in the CytoScan assay?
The tested sources of human gDNA used in the CytoScan Assay are blood and cell line samples.
The genomic DNA (gDNA) sample must be double-stranded, not degraded and free of any contaminants (e.g., PCR inhibitors and other human/non-human gDNA). The recommended starting amount is 250 ng (5 µL with 50 ng/µL) dissolved in low EDTA TE buffer. High EDTA concentration may negatively impact the downstream enzymatic reactions. We recommend running the gDNA samples on a 0.8-1% agarose gel for side-by side comparison with a control DNA (included in the kit). High-quality genomic DNA will run as a major band at approximately 10-20 kb on the gel.
For more detailed information please refer to the CytoScan Assay User Guide, page 9, Genomic DNA general requirements and recommendations section .
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which type of freezer should be used to store CytoScan Reagent kit components that require -20 degrees C?
The reagents that require storage at -20 degrees C must not be stored in a frost-free freezer. The activity of the enzymes will be decreased.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the maximum number of freeze/thaw cycles recommended for the CytoScan Reagent kit?
The CytoScan Reagent kit has been validated for less than or equal to 5 freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why does the protocol state to prepare a fragmentation master mix enough for 24 samples even if a smaller number of samples is processed?
It is critical to have correct size of the fragments for an optimal hybridization. The fragmentation reagent is very viscous and it is easier to get a correct proportion of enzyme when a larger volume is used. The recommendation is to always prepare a master mix for 24 samples even if a smaller number of samples are processed.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which thermal cyclers are compatible with the CytoScan Assay protocol?
Thermal cyclers with the following specifications are compatible with the CytoScan Assay protocol:
  • Capable of holding 200 µL volume and 96-well plate
  • Heat block capable of holding temperature of 4-99.9°C
  • Temperature accuracy of +/- 0.25°C (at 35-99.9°C)
  • Average heating and cooling rate of 2.6°C /second
  • Thermal uniformity of +/- 0.5°C
The following PCR instruments have been verified for use in the CytoScan Assay (newer thermal cyclers may use simulation mode to mimic these settings):
  • GeneAmp PCR System 9700 Gold/Silver
  • Applied Biosystems Veriti 96-Well Thermal Cycler (Cat. No. 4375786)
  • Applied Biosystems Veriti Dx 96-Well Thermal Cycler (Cat. No. 4452300)
  • Bio-Rad DNA Engine PTC-200 Thermal Cycler (Calculated Temperature Mode)
  • Eppendorf Mastercycler Pro S Thermal Cycler (Safe Temperature Mode) (Cat. No. 950030020 and Cat. No. 950040025).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which are the possible stopping points in the CytoScan Reagent kit assay?
The plate can be tightly sealed and stored at -20 degrees C after all steps of the assay. The plate can be left over night at 4 degrees C after the PCR reaction and labeling reaction have been completed.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How should the thermal cycler settings be programmed for use with the CytoScan 750 array?
  Applied Biosystems Veriti 96-well Bio-Rad DNA Engine PTC-200 Eppendorf Mastercycler Pro S
Lid temperature 103 degrees C 103 degrees C 103 degrees C
Temperature mode N/A Calculated Safe
TSP heated lid N/A N/A Yes
Switch off lid at a low block temperature N/A N/A No
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When using ChAS, is it important for someone to back up their data?
Out of the six data files mentioned in the preceding question, it is important to backup and archive the ARR, CEL, CYCHP, and CHPCAR files at a minimum. This will allow someone to maintain the ability to either reanalyze from the CEL file or re-visualize the results using the CYCHP/CHPCAR files.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What data files are produced during assay and analysis process when using ChAS?
A total of six files are produced that are key to the process:
ARR file: this file includes sample information.
AUDIT file: this file is a log of the sample history.
DAT file: this file is the raw data from the scanner.
CEL file: this file is the gridded and processed data.
CYCHP file: this file is the output of ChAS and contains all of the analysis data.
CHPCAR file: this file stores user-annotated calls, interpretations, and modifications made to CHP file segment data (ChAS 2.0 and higher).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the mosaic call determination in ChAS?
The algorithm is designed to detect only mosaicism between ~30-70% for CNs between 1 and 3 for regions on the order of 5,000 markers in size or larger. The endpoint location of mosaic segments is less precise than the CN segmentation, with endpoint variation within 500 markers being typical for segments of 5,000 markers or larger. Some regions of full integer CN 1 or 3 below 5,000 markers in length may be incorrectly called as mosaic segments. Some regions of CN below 1 or above 3, mosaic or otherwise, less than 5,000 markers in length may be incorrectly called as mosaic segments.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the library file location for ChAS?
The library file location for ChAS 2.1 and above is C:\Affymetrix\ChAS\Library.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I unzip the array library file if I have to install it manually?
Right click the zip folder and click extract all. They can also use an already installed software that unzips files (e.g., Winzip, 7-zip, or any other equivalent software). Browse to the location to save the folder and click Extract. Go to the folder to find the files.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How does someone export LogRatios for the whole genome?
They can export the Whole Genome View as a PNG file. To do this, click on File > Export window PNG.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are there additional databases to refer to that will help in data analysis for the CytoScan HD Array?
Yes, refer to the CAGdb database, ICCG, and DECIPHER for additional public databases that may assist in data analysis, and for which we do not have a direct link within ChAS.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What samples are in the Reference Model file in the CytoScan HD Array and CytoScan 750K Array set?
The Reference Model file in the CytoScan HD Array and CytoScan 750K Array set includes 380 microarrays, which were run as part of a larger set of microarrays by nine operators. These operators processed ~48 unique samples in two rounds each, with random placement of sample DNAs across the PCR plates and with random use of reagents and instruments. The source DNA includes the following:
284 HapMap samples including at least one replicate of each of 270 HapMap samples:
90 from each of the Yoruban, Asian, and Caucasian ethnic groups, from cell-line derived DNAs from the Coriell Institute of Medical Research
96 DNA samples from blood of phenotypically healthy male and female individuals obtained from BioServe Biotechnologies
The CytoScan HD Array samples that were used to create the Reference Model file were chosen to be run by different operators and with different kits and reagents, while still covering all the HapMap cell line ethnic groups, plus the normal bloods of both sexes.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which genotype is presented in each allele difference track?
AA markers would be distributed about +1.
AB markers would be distributed about 0.
BB markers would be distributed about -1.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the recommended graph settings for data interpretation in ChAS?
We recommend changing the weighted log2 ratio graph from the default to a minimum at -1.5 to a maximum at 1.5, using data type as points. We recommend changing the allele peaks graph from the default to a minimum at -2 to a maximum at 2, using data type as points. The other graphs can stay with the default values.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are individual genotypes generated from the CytoScan arrays?
Yes, genotypes can be generated in ChAS. The software uses BRLMM for genotyping, which uses a Bayesian model based on prior clusters. This product is not intended for genome-wide association studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For CytoScan arrays, what is Waviness-SD?
Waviness-SD is a global measure of variation of microarray probes that is insensitive to short-range variation and focuses on long-range variation. Based on an empirical testing dataset, array data with Waviness-SD >0.12 has either sample or processing batch effects that will reduce the quality of the copy number calls. Elevated Waviness-SD is not always an indication of too much noise. Elevated Waviness with good MAPD and SNPQC metrics can occur in samples with many copy number changes or very large regions of change. It is therefore advised to check the data when observing elevated Waviness with good MAPD and SNPQC. The Waviness-SD metric is applicable to blood and cell line data. The Waviness-SD metric is not intended for alternative sample types such as solid tumor or FFPE samples in which the results may vary as a result of the biological complexity. For these sample types, it is recommended to use the ndwavinessSD.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the definitions of the array QC metrics terms "MAPD" and "SNPQC"; what does each metric measure and how are they used in assessing array performance?
MAPD: Median of the Absolute values of all Pairwise Differences is a per-microarray estimate of variability, like standard deviation (SD) or interquartile range (IQR). It measures the variability in log2 ratios by looking at the pair difference of all probes and taking a median value. The effect of an occasional big difference in log2 ratios between probes is removed by taking a median value and not a mean. This variability can come from different sources: Intrinsic variability in the starting material, hybridization cocktail preparation, microarray, or scanner.
Apparent variability induced by the fact that the reference may have systematic differences from the sample on this microarray. Regardless of the source of variability, increased variability decreases the quality of the CN calls. A high MAPD can be attributed to any of the above factors and indicates that CN calls may be inaccurate, leading to a higher false positive/negative rate.
SNPQC: This is a measure of how well genotype alleles are resolved in the microarray data. In other words, it estimates the distributions of homozygous AA, heterozygous AB, and homozygous BB alleles and calculates the distance between them. The better the separation of these distributions, the better the ability to identify a genotype based on its cluster position. The larger the difference between the peaks and the troughs, the better the resolution of homozygotes and heterozygotes and the higher the SNPQC metric is. If the three peaks are not well resolved, the difference between peaks and troughs will be low, resulting in a lower SNPQC value. A low SNPQC value indicates that quality of the SNP allele data is compromised, due to higher noise within the array, which compromises the overall quality and clarity of results.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What software is available for analysis for genoytping and copy number analysis?
All genotyping and copy number analysis is done with ChAS. CytoScan Cytogenetics Suite is not intended for genome-wide association studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What file sizes are associated with CytoScan HD Array?
The CEL file is ~66 MB, and the CYCHP file is ~119 MB.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the correct fluidics script for CytoScan HD Array?
The correct fluidics script is CytoScanHD_Array_450.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long does CytoScan HD Array take to scan?
Each array takes ~32 minutes to scan.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How many single-nucleotide polymorphism (SNP) and copy number (CN) probes are included on the array?
CytoScan HD Array includes 750,000 genotype-able SNPs and 1.9 million non-polymorphic probes.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the array content for the CytoScan HD Array?
CytoScan HD Array covers both constitutional and cancer genes with:
Overall intragenic coverage at 1 marker/880 bases
ClinGen (previously CCG and ISCA) constitutional coverage at 1 marker/384 bases
Complete cancer gene coverage at 1 marker/553 bases
14,000 OMIM genes at 1 marker/723 bases
>36,000 RefSeq genes at 1 marker/880 bases
Backbone (non-gene) coverage at 1 marker/1,737 bases across genome for breakpoints Overall (gene and non-gene backbone) coverage at 1 marker/1,148 bases
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the array format for the CytoScan HD Array?
CytoScan HD Array uses a 49 format and a 5-µm feature size.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are there any shutdowns run after bleaching the fluidics station when working with the CytoScan arrays?
Yes, it is recommended to run two water shutdown protocols after bleaching the fluidics station.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long can CytoScan arrays filled with holding buffer be stored before scanning?
After the washing and staining step, arrays can be stored for up to 24 hrs at 4 degrees C before scanning.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
After hybridization, how long can CytoScan arrays be stored before washing and staining?
Arrays must be put onto the fluidics station immediately after removal from the hybridization oven. Do not remove arrays from the oven you are ready to wash them.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the hybridization time for CytoScan arrays?
The hybridization time is 16-18 hrs. If there are a limited number of fluidics stations, the hybridization process can be staggered by two hrs. For example, if there is only one fluidics station and eight arrays, the hybridization for four arrays can be started at one time, and then the hybridization for the other four arrays started two hrs later. After 16 hrs, the first four arrays can be placed on the fluidics station, and then the next four arrays placed on the fluidics station two hrs later. Thus, all eight samples will have hybridized for 16 hrs. Another option is to wash the first set of samples at 16 hrs and the second set at 18 hrs. It is not recommended to stagger more than three wash/stain cycles in an eight-hour workday. Please consult technical support at techsupport@thermofisher.com for more details on the hybridization process.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long can the labeled products be stored at -20 degrees C?
The labeled products can be stored at -20 degrees C for no more than 10 days.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can cause under-fragmentation for the CytoScan assay?
Common causes of under-fragmentation include improper storage or handling of the enzyme, an incorrect volume of enzyme used based on the unit activity, and improper mixing of fragmentation master mix. See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more information about under-fragmentation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can cause over-fragmentation for the CytoScan assay?
Common causes of over-fragmentation include excess enzyme due to pipetting errors or an incorrect volume of enzyme used based on the unit activity. In addition, warming up of the assembled reaction prior to initiating the 37 degrees C incubation can lead to over-fragmentation. See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more information about over-fragmentation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Do the samples for the CytoScan Assay need to be run on a gel and the Bioanalyzer during fragmentation QC?
No, the routine QC includes running samples only on a gel. Some samples will be saved for additional analysis on the Bioanalyzer, should that become necessary.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What should be done if a refrigerated centrifuge is not available for the fragmentation step in the CytoScan Assay?
A refrigerated centrifuge is highly recommended. If one is not available, a plastic plate rack that has been stored at -20 degrees C may be used. It is recommended to keep the samples chilled and work quickly prior to initiating the incubation at 37 degrees C.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the purpose of the fragmentation step of the CytoScan Assay?
Probes on the array are 25-mers. The fragmentation step reduces the purified PCR product size (150- 2,000 bp) to an even smaller size range (25-125 bp), making them more optimal for hybridization to the array probes. Ideally, 25 bp-sized PCR products hybridize to 25-mer probes on the array; however, optimal hybridization can be achieved with 25 to 125 bp-sized PCR products.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long can the purified PCR products be stored at -20 degrees C?
The purified PCR products can be stored at -15 to -25 degrees C for 10 days.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I stop after the PCR process and continue the next day with the purification step for the CytoScan Assay?
Yes, the purified PCR products can be stored at -15 to -25 degrees C, and one can continue with the purification step later.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Assay, what is the role of purified PCR product?
The purification process is required to remove all the non-amplified DNA after the PCR process.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the typical yield range from PCR product purification?
The minimum recommended yield is 2.5 µg/µL for a sample. Yields can range from 3.0-4.5 µg/µL, and the average yield for seven or more samples processed in a run (not including the negative control) should be greater than 3.0 µg/µL. If the average yield is below this, consult the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan assay, can a different magnetic rack be used instead of the recommended MagnaRack magnetic stand?
Yes, the assay is currently validated for the following magnetic racks:
DynaMag-2 Magnet (Cat. No. 123-21D)
MagnaRack (Cat. No. CS15000)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan assay, is there a plate purification option for the PCR product purification step?
No, only the single-tube purification process is validated with the assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What should I do if I see a smear in their negative control on their PCR gel when performing the CytoScan assay?
A smear in the negative control indicates there has been a contamination. If the negative control band is a light smear, it could be a low-level environmental bacterial contaminant introduced through plastic surfaces. This has shown not to have a material impact on the CytoScan HD data. If a bright band or a smaller smear is seen, then that could be a real cross-contaminant from a sample, and the samples would need to be checked for contamination. If a smear is seen in the negative control well, you should re-run the negative control to verify this was not a result of sample bleed-over from gel loading.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan assay, does the PCR have to be set up in a separate pre-PCR clean room?
Using two separate rooms greatly reduces the risk of sample contamination by previously amplified PCR products. If only one room is available, designate one area of the room as the pre-PCR clean area and a separate area as the post-PCR clean area. If a one-room configuration is being used, it is highly recommended to use a laminar flow cabinet for the pre-PCR clean area. See the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more details about the recommended laboratory setup.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
In a hypothetical CytoScan assay failure in which PCR fragment size distribution observed on the PCR gel is significantly greater than 2,000 bp DNA smear, what processes in the assay could play a role in the failure?
The digestion step could be root cause. If the reaction did not go to completion, there may be, on average, longer fragments at the end of the reaction. These longer fragments then pass on to ligation and PCR reactions.
Insufficient amounts of PCR primer were added to facilitate suppression PCR. The primer concentration shifts the PCR reaction equilibrium toward larger fragment distributions, in this case, by increasingly unfolding (linearizing) the stem-loop structure at increasing primer concentration when the same adaptor ends hybridize.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When performing the CytoScan Assay, what can cause faint or absent bands on the PCR gel?
Several things can cause this problem. To troubleshoot this problem, first determine if the positive control worked properly. Common reasons for this failure include incomplete digestion of genomic DNA or inefficient ligation of adaptors, ligation samples that are not properly diluted or mixed, and degraded DNA (if only the positive control worked). See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038) for more information
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long can the ligated products for the CytoScan assay be stored at -20 degrees C?
A plate can be frozen at -15 to -25 degrees C for up to one week.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long can the ligated products for the CytoScan assay be stored on the thermal cycler?
A plate can be held in thermal cycler at 4 degrees C for up to 60 hrs.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What happens during the digestion and ligation steps of the CytoScan assay?
The DNA is digested with Nsp I restriction endonuclease and ligated to adaptors that recognize the cohesive four-base pair (bp) overhangs. All fragments resulting from restriction enzyme digestion, regardless of size, are substrates for adaptor ligation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the 10 CytoScan assay processing steps listed in order?
The 10 assay processing steps, as listed in the CytoScan Assay User Manual (Cat. No. 703038) are the following:
1. Genomic DNA Preparation
2. NSP1 Restriction Enzyme Digestion
3. Ligation
4. PCR
5. PCR Product Purification
6. Quantitation
7. Fragmentation
8. Labeling
9. Hybridization
10. Wash, Stain, Scanning
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why do the pre- and post-PCR areas need to be separated?
The post-PCR lab contains amplicons, whereas the pre-PCR area should not. These amplicons can be inadvertently transferred back to the pre-PCR area, thus contaminating the lab and posing a contamination threat to all reactions set up in the pre-PCR lab. Because all samples are amplified using the same primers and adaptors, contaminating DNA can also be amplified.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the purpose of including a positive and negative genomic DNA control in each CytoScan assay, and what would be the passing and failing results for each control type?
Positive control: This sample has prior proven performance and represents a pristine sample, meeting all genomic DNA input requirements. The performance of this sample helps to assess the quality of the assay steps and final product. In troubleshooting cases, performance of this control sample versus that of non-control samples can help to parse out possible assay failure culprits.
Negative control: For this assay, the negative control is simply reduced EDTA TE Buffer and acts as a control for the process up to the PCR quantitation and PCR gel. If no smear is seen on the PCR gel, it is considered passing, indicating that there is no DNA contamination in any of the reagents that could lead to amplification of other targets. A failing result will have a smear in the PCR gel. Note that the presence of PCR primer dimers is expected.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I buy just the reagents or arrays alone instead of the CytoScan Reagent Kit?
No, the reagents and arrays are sold as bundles for 24 reactions (Cat. No. 901835)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How is the CytoScan assay different from the assay used with Affymetrix Genome-Wide Human SNP Array 6.0?
The CytoScan assay has significantly fewer pipetting steps and requires less hands-on time than the Genome-Wide Human SNP 6.0 assay. The CytoScan assay uses only the Nsp I restriction enzyme and has been optimized for cytogenetics applications. The CytoScan assay is not intended for genome-wide association studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the stopping points in the CytoScan Reagent Kit assay?
After any stage in the assay (except digestion), samples can be stored at -20 degrees C if one is not proceeding directly to the next step. However, once a stage has been initiated, it must be completed before storage of the samples.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Reagent Kit, does the expiration date apply to the date of opening or date of manufacturing?
The expiration date applies to the date of manufacturing. For CytoScan Reagent Kit, the reagents can be used for up to one year from the date of manufacturing.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can reagents from Affymetrix SNP 6 Core Reagent Kit be used in the CytoScan assay?
No. The CytoScan assay has been optimized for performance with only CytoScan Reagent Kit.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can different sub kits from different lots of the modules from the CytoScan assay kit be used for a particular experiment?
It is highly recommended not to mix different sub kit lots from different modules because functional testing is done based on sub kits from the same lot.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the fill volumes of the reagents in CytoScan Reagent Kit?
Only the volume that is indicated on the label can be guaranteed. Since our assurance guarantee is only provided for the label volume, it is advised not to plan experiments or standard operating procedures based on potential overage volumes.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How many samples should be processed at a time using the CytoScan assay?
It is recommended to run no more than 24 samples at a time. The assay is a four-day protocol; however, the technician can use an optional three-day protocol after becoming comfortable with the assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the recommended genomic DNA extraction methods used for the CytoScan assay?
Both blood and cell line sample sources have been tested with the CytoScan assay.
Methods that include boiling or strong denaturants are not acceptable because the DNA would be rendered single-stranded. Genomic DNA extracted using the following methods have been tested at ThermoFisher Scientific:
QIAGEN - Gentra Puregene Kit
5 PRIME - PerfectPure DNA Blood Kit
The CytoScan assay requires genomic DNA concentration >50 ng/µL. Therefore, the elution volumes for each of the kits will need to be adjusted accordingly to achieve the desired concentration.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the general genomic DNA requirements for the CytoScan assay?
DNA must be double-stranded genomic DNA.
DNA must be free of PCR inhibitors.
DNA must not be contaminated with other human genomic DNA sources or with genomic DNA from other organisms.
DNA must not be degraded.
DNA should have an A260/A280 between 1.7-2.1 (numeric rounding allowed).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What sample types are validated on the CytoScan assay?
The CytoScan assay has been validated with blood and cell line samples.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which magnetic racks are supported for the CytoScan assay?
The use of the following magnetic racks is supported:
-DynaMag-2 Magnet (Cat. No. 123-21D)
-MagnaRack (Cat. No. CS15000)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can electronic pipettes be used in the CytoScan assay instead of multichannel manual pipettes?
Pipetting viscous solutions using electronic pipettes may be very challenging and result in ineffective pipetting that could contribute to overall poor QC values. Therefore, the use of electronic pipettes is not recommended.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Am I required to have all equipment current regarding calibration and maintenance?
For successful execution of the CytoScan assay, it is important that equipment be properly maintained and calibrated per the manufacturers specifications. Please be sure that the centrifuges, pipettes, thermal cyclers, and ThermoFisher Scientific equipment, including the 645 hybridization oven, fluidics station(s), and scanner, have had their recommended maintenance prior to running the assay. For lab equipment, it is typically a yearly calibration. For ThermoFisher Scientific equipment, there is typically a yearly preventative maintenance.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is required for processing and viewing CytoScan array CEL files?
CytoScan array CEL files are processed and analyzed in ChAS. This is a free download from the ThermoFisher Scientific web site. For processing and viewing the CEL files, a 64-bit computer with a minimum of 8 GB of RAM, is required. The recommended system specifications are included in the Chromosome Analysis Suite User Manual (Cat. No. 702943; http://assets.thermofisher.com/TFS-Assets/LSG/manuals/ChAS_Manual.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How can I tell the difference between a silver and aluminum thermal cycler block?
Look at the serial number on the block. An “A” in the serial number indicates the block is aluminum; an “S” indicates it is silver. In addition, an aluminum block has a honeycomb appearance between the wells, whereas a silver block is smooth.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What thermal cycler can be used in for the CytoScan assay?
Applied Biosystems GeneAmp PCR System 9700 (silver or gold-plated silver block only; do not use an aluminum block)
Applied Biosystems Veriti 96-Well Thermal Cycler
Bio-Rad DNA Engine PTC-200 Thermal Cycler
Eppendorf Mastercycler Pro S Thermal Cycler
Applied Biosystems 2720 (pre-PCR room only)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What Thermo Fisher Scientific equipment is required for the CytoScan assay?
The CytoScan assay requires either GeneChip Scanner 3000 7G System with Workstation and AutoLoader or GeneChip System 3000Dx v.2 and a GeneChip Hybridization Oven 645.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is CytoScan Cytogenetics Suite?
CytoScan Cytogenetics Suite consists of CytoScan arrays and CytoScan Reagent Kit, GeneChip Command Console Software (AGCC), and Chromosome Analysis Suite (ChAS). It enables the performance of high-resolution, genome-wide DNA copy number analysis and also provides genotyping information for the detection of copy neutral loss/absence of heterozygosity (LOH/AOH), which can be used to detect uniparental isodisomy (UPD). The combination of high-resolution DNA copy number data and the ability to detect gains, losses, and UPDs on a single array makes CytoScan Cytogenetics Suite ideal for cytogenetics studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan 750K Array, is it important to back up my data in ChAS?
Out of the six data files (ARR, AUDIT, DAT, CEL, CYCHP and CHPCAR), it is important to backup and archive the ARR, CEL, CYCHP and CHPCAR file at a minimum. This will allow you to maintain the ability to either reanalyze from the CEL file, or re-visualize the results using the CYCHP/CHPCAR files.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan 750K Array, what data files are produced during assay and analysis process?
A total of 6 files are produced that are key to the process:
ARR file - This file includes sample information
AUDIT file - This file is a log of the sample history
DAT file - This file is the raw data from the scanner
CEL file - This file is the gridded and processed data
CHPCAR file - This file stores user-annotated calls, interpretations, and modifications made to CHP file segment data
CYCHP file - This file is the output of ChAS and contains all of the analysis data
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are individual genotypes generated on the CytoScan 750K Array?
Yes, genotypes can be generated in the Chromosome Analysis Suite (ChAS 3.3) Software. The software uses BRLMM for genotyping, which uses a Bayesian model based on prior clusters. This product is not intended for genome-wide association studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can cause waviness SD failures when using the CytoScan Assay?
Waviness SD failures can be caused by degraded DNA, an incompatible sample type, or a sample-specific effect. See the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) and the Chromosome Analysis Suite User Manual (Cat. No. 702943) for more information.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can cause MAPD failures when using the CyotScan Assay?
MAPD failures can be caused by assay drift due to variation in assay execution or over fragmentation. See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more information.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can cause SNPQC failures when using the CytoScan Assay?
SNPQC failures can be caused by contamination of samples, equipment, or reagents. Other possible causes include over or under fragmentation of samples or a hybridization oven that is out of calibration. See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038) for more information (https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What type of water should be used on the fluidics stations?
Always use deionized (DI) water on the fluidics stations for all protocols, including the shut down and bleach protocols.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Assay, what can cause under-fragmentation?
Common causes of under-fragmentation include improper storage or handling of the enzyme, an incorrect volume of enzyme used based on the unit activity, and improper mixing of fragmentation master mix. See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more information about under-fragmentation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Assay, what can cause over-fragmentation?
Common causes of over-fragmentation include excess enzyme due to pipetting errors or an incorrect volume of enzyme used based on the unit activity. In addition, warming up of the assembled reaction prior to initiating the 37 degrees C incubation can lead to over fragmentation. See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more information about over-fragmentation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Do I need to run the samples on a gel and the Bioanalyzer during fragmentation QC?
No, the routine QC includes running samples only on a gel. You will save some samples for analysis in case you need them for additional analysis on the Bioanalyzer.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What should I do if I do not have a refrigerated centrifuge for the fragmentation step when working with a CytoScan array?
A refrigerated centrifuge is highly recommended, but if one is not available, you may use a plastic plate rack that has been stored at -20 degrees C. It is recommended that you keep the samples chilled and work quickly prior to initiating the incubation at 37 degrees C. See the CytoScan Assay and Data Analysis Training Video (www.thermofisher.com/us/en/home/life-science/microarray-analysis/microarray-analysis-learning-center/microarray-analysis-training.html) for more details.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Assay, what is the typical yield range from PCR product purification?
The minimum yield recommended is 2.5 µg/µL. Yields can range from 3.0-4.5 µg/µL and the average yield for seven or more samples processed in a run (not including the negative control) should be greater than 3.0 µg/µL. If the average yield is below this, consult the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038, https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I use a different magnetic rack instead of the recommended MagnaRack magnetic stand for the CytoScan Assay?
No, the Magna Rack is the only magnetic rack that is validated with the assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Assay, what can cause faint or absent bands on the PCR gel?
Several things can cause this problem. The first step in troubleshooting this problem is to determine if the positive control worked. Common reasons for this failure include incomplete digestion of genomic DNA or inefficient ligation of adaptors, ligation samples that are not properly diluted or mixed, and degraded DNA (if only the positive control worked). See the troubleshooting section of the CytoScan Assay User Manual (Cat. No. 703038; https://tools.thermofisher.com/content/sfs/manuals/cytoscan_assay_user_manual.pdf) for more information.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan Assay, what happens during the digestion and ligation steps?
The DNA is digested with Nsp I restriction endonuclease and ligated to adaptors that recognize the cohesive four base pair overhangs. All fragments resulting from restriction enzyme digestion, regardless of size, are substrates for adaptor ligation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How is the CytoScan Assay different from the assay used with the Affymetrix Genome-Wide Human SNP Array 6.0?
The CytoScan Assay has significantly fewer pipetting steps and requires less hands-on time than the Affymetrix Genome-Wide Human SNP 6.0 Assay. The CytoScan Assay uses only the Nsp I restriction enzyme and has been optimized for cytogenetics applications. The CytoScan Assay is not intended for genome-wide association studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the stopping points in the CytoScan Assay?
After any stage in the assay (including fragmentation) you can store the samples at -20 degrees C if you are not proceeding directly to the next step. However, once you have initiated a stage, you must complete it before storage of the samples.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For the CytoScan assay, does the expiration date apply to the date of opening or date of manufacturing?
The expiration date applies to the date of manufacturing. For the CytoScan Reagent Kit, it can be used for up to one year from the date of manufacturing.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I use reagents from the Affymetrix SNP 6 Core Reagent Kit in the CytoScan Assay?
No. The CytoScan Assay has been optimized for performance with the CytoScan Reagent Kit, which is therefore required.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What other reagents are required for the CytoScan Assay?
The CloneTech Titanium Taq PCR Kit (300 reactions, Cat. No. 639240 or 400 reactions, Cat. No. 639243) and absolute ethanol (Sigma Cat. No. 459844) are also required. All other reagents are included in the CytoScan Reagent Kit.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the throughput for the CytoScan Assay?
We recommend running no more than 24 samples at a time. The assay is a four-day protocol; however, the technician can use an optional three-day protocol after becoming comfortable with the assay
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How many freeze/thaw cycles are validated for the CytoScan Assay 24-Sample Kit?
The CytoScan Reagent Kit (Cat. No. 901808) is validated for four freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Is it required that all my equipment is current on its calibration and maintenance status?
For successful execution of the CytoScan Assay, it is important that your equipment is properly maintained and calibrated per manufacturers' specifications. Please assure that your centrifuges, pipettes, thermal cyclers, and Affymetrix equipment, including the 645 Hybridization Oven, Fluidics Station(s), and Scanner, have had their last recommended maintenance prior to running the assay. For lab equipment, it is typically a yearly calibration. For the Affymetrix equipment, there is typically a yearly preventative maintenance.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How can you tell the difference between a silver and aluminum thermal cycler block?
Look at the serial number on the block. An “A” in the serial number indicates the block is aluminum; an “S” indicates it is silver. Also, an aluminum block has a honeycomb appearance between the wells, whereas a silver block is smooth.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What thermal cycler can be used for the CytoScan Assay?
The ABI 9700 (silver or gold-plated silver block only; do not use an aluminum block) and the ABI 2720 (pre-PCR room only) thermal cyclers have been validated for the CytoScan Assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the CytoScan Assay?
The CytoScan Assay, along with the CytoScan 750K Arrays, the GeneChip Command Console Software (AGCC), and the Affymetrix Chromosome Analysis Suite (ChAS) Software, enable you to perform high-resolution genome-wide DNA copy number analysis. This Affymetrix solution for cytogenetics also provides genotyping information, enabling detection of copy neutral loss of heterozygosity (LOH), which can be used to detect uniparental isodisomy (isodisomic UPD). The combined high-resolution DNA copy number data and the ability to detect gains, losses, and UPDs on a single array makes the CytoScan 750K Solution ideal for next-generation cytogenetics studies.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When would we choose normal diploid analysis over single-sample analysis?
The normal diploid analysis for a CytoScan HD and 750K array is recommended for cancer samples in which greater than 50% of the genome is likely to be rearranged. This analysis automatically determines the normal diploid regions and normalizes the rest of the samples based on those regions, resulting in properly centered data. The normal diploid analysis is NOT recommended for CytoScan Optima Array.
For samples run through the normal single sample analysis, the following QC metrics are recommended:
MAPD less than 0.25
SNPQC OR ndSNPQC greater than or equal to 15
Waviness-SD OR ndwavinessSD less than 0.12
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What samples are in the CytoScan HD and 750K reference model files?
The CytoScan HD and 750K reference model files are comprised of the same set of samples and include 380 microarrays that were run as part of a larger set of microarrays by nine operators processing ~48 unique samples in two rounds each, with randomization of the placement of DNA samples across the PCR plates and randomization of the reagents and instruments used. The source DNA includes
284 HapMap samples, including at least one replicate of each of 270 HapMap samples: 90 from each of the Yoruban, Asian, and Caucasian ethnic groups from cell line-derived DNA from the Coriell Institute of Medical Research
96 DNA samples from blood of phenotypically healthy male and female individuals obtained from BioServe Biotechnologies
The samples in this set were chosen to have been run by different operators and with different kits and reagents while still covering all the HapMap cell line ethnic groups, plus the normal blood samples of both sexes.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are individual genotypes generated on CytoScan arrays?
Yes, genotypes can be generated in ChAS 3.1 and above. The software uses BRLMM for genotyping, which uses a Bayesian model based on prior clusters. This product is not intended for genome-wide association studies
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the array QC metrics for the CytoScan arrays?
CytoScan HD and 750K arrays use the following QC metrics:
SNPQC greater than or equal to 15
MAPD less than or equal to 0.25
Waviness-SD less than or equal to 0.12
CytoScan Optima Array uses the following QC metrics:
SNPQC greater than or equal to 8.5
MAPD less than or equal to 0.29
Waviness-SD less than or equal to 0.12
For CytoScan HD Suite and CytoScan 750K Suite, QC metrics have been fine-tuned for blood- derived constitutional samples. The SNPQC and Waviness-SD metrics are based on an assumption of a relatively normal diploid genome for which the majority of the genome is not mosaic. For cancer- related samples, the baseline assumption pertaining to constitutional samples is violated with regard to aberration frequency and high levels of mosaicism, which will likely trigger the SNPQC and Waviness-SD metrics to fail. Only the MAPD metric should be considered for non-constitutional samples. A failure of any one of these metrics for constitutional blood samples is a failure for that array result. There is no direct correlation between the absolute passing numeric value for any one of the metrics and the quality of a sample.
For CytoScan Optima Suite, QC metrics have been fine-tuned for amniocytes, chrionic villi, cultured cells, POC, and blood samples. The SNPQC and Waviness-SD are based on an assumption of a relatively normal diploid genome.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is waviness standard deviation (Waviness-SD), and how should it be used?
Waviness-SD is a global measure of variation of microarray probes that is insensitive to short-range variation and focuses on long-range variation. Based on an empirical testing dataset, we have determined that array data with Waviness-SD >0.12 has either sample or processing batch effects that will reduce the quality of the copy number calls. Elevated Waviness-SD is not always an indication of too much noise. Elevated waviness with good MAPD and SNPQC metrics can occur in samples with many copy number changes. Therefore, it is advised to check the data when observing elevated waviness with good MAPD and SNPQC. The Waviness-SD metric is applicable to constitutional blood and cell line data. The Waviness-SD metric is not intended for alternative sample types such as cancer samples in which the results may vary as a result of the biological complexity. For these sample types, it is recommended to use the ndwavinessSD.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is SNP quality control (SNPQC), and how should it be used?
SNPQC is a measure of how well genotype alleles are resolved in the microarray data. In other words, it estimates the distributions of homozygous AA, heterozygous AB, and homozygous BB alleles and calculates the distance between them. The better the separation of these distributions, the better the ability to identify a genotype based on its cluster position. A low SNPQC value indicates that the quality of the SNP allele data is compromised due to higher noise within the array, which compromises the overall quality and clarity of results.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is median of the absolute values of all pairwise differences (MAPD), and how should it be used?
MAPD is a per-microarray estimate of variability, like standard deviation (SD) or interquartile range (IQR). It measures the variability in log2 ratios by looking at the pair difference of all probes and taking a median value. The effect of an occasional big difference in log2 ratios between probes is removed by taking a median value and not a mean. This variability can come from different sources:
-Intrinsic variability in the starting material, hybridization cocktail preparation, microarray, or scanner
-Apparent variability induced by the fact that the reference may have systematic differences from the sample on this microarray
Regardless of the source of variability, increased variability decreases the quality of the CN calls. A high MAPD can be attributed to any of the above factors and indicates that CN calls may be inaccurate, leading to a higher false positive/negative rate.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What file sizes are associated with the CytoScan HD Array?
The CEL file is approximately 66 MB, and the CYCHP file is approximately 120 MB.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How many single nucleotide polymorphism (SNP) and copy number (CN) probes are included on the CytoScan HD Array?
CytoScan HD Array includes a total of approximately 6.5 million probes: 1 probe per allele in triplicate for 750,000 SNPs probes and 1.9 million non-polymorphic probes.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the CytoScan HD Array content?
-CytoScan HD Array covers both constitutional and cancer genes with Overall intragenic coverage at 1 marker/880 bases
-ClinGen (ISCA) constitutional coverage at 1 marker/384 bases
-Complete cancer gene coverage at 1 marker/553 bases
-12,000 OMIM genes at 1 marker/659 bases
->36,000 RefSeq genes at 1 marker/880 bases
-Backbone (non-gene) coverage at 1 marker/1,737 bases across the genome for breakpoints
-Overall (gene and non-gene backbone) coverage at 1 marker/1,148 bases
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the Median Absolute Pairwise Difference (MAPD) metric?
MAPD is the recommended method of assessing noise in the SNP Array 6.0, Cytoscan HD and Cytoscan DX copy number data. The log2 distance between neighbors is calculated for all pairs and then the median is taken. This represents the typical distance between marker pairs with respect to log2 ratios. The higher the MAPD, the greater the noise between the array of interest and the reference set.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.