Axiom™ 2.0 384HT Reagent Kit - FAQs
Axiom™ 2.0 384HT Reagent Kit - FAQs
View additional product information for Axiom™ 2.0 384HT Reagent Kit - FAQs (902245)
1 product FAQs found
Why do I need to use the Control Oligo B2 with my assay?
Control Oligo B2 (3 nM) is a pre-labeled DNA spike-in control required for hybridization to the control probes on the array utilized for grid alignment by AGCC. A failure to include Control Oligo B2 in the hybridization cocktail will result in the inability of the software to apply a grid over the scanned image, leading to the unrecoverable loss of sample data from the array. The absence of the Control Oligo B2 also voids the customer's ability to submit an array replacement request.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.