Axiom™ Starter Kit for NIMBUS
Applied Biosystems™
Axiom™ Starter Kit for NIMBUS
The Axiom™ Starter Kit for NIMBUS™ products includes additional equipment and parts necessary to set up the NIMBUS Target PreparationMás información
Have Questions?
Número de catálogo 902365
Precio (MXN)
-
The Axiom™ Starter Kit for NIMBUS™ products includes additional equipment and parts necessary to set up the NIMBUS Target Preparation Instrument for Axiom Genotyping Solution.
We have partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. NIMBUS Target Preparation Instrument is industry-recognized for its high-end manufacturing and superior pipetting performance at an affordable price.
Axiom 96-sample automated target preparation
We have automated the target preparation steps for the Axiom 2.0 assay, including DNA amplification, fragmentation, purification, resuspension, hybridization preparation, and stain preparation for the GeneTitan™ MC Instrument.
The Axiom target preparation on the automated liquid-handling workstation helps minimize run-to-run variability and the labor burden associated with complex manual pipetting, helping to improve test reproducibility and laboratory efficiency.
Target preparation for Axiom Genotyping Solution is now effortless with Axiom 96-sample automated target preparation, which offers:
• Small footprint, sleek design, and industry-recognized engineering
• Advanced pipetting technology for precise yet flexible liquid handling
• Workflow designed to accommodate 96 samples
• Laptop with easy-to-use software interface including visual cues for each assay step
Related Links
NIMBUS™ Target Preparation Instrument
Axiom™ Consumables Kit for NIMBUS
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Para utilizar con (aplicación)Microarray Analysis
Para utilizar con (equipo)NIMBUS Target Preparation Instrument
Cantidad1 kit
TipoStarter Kit for NIMBUS
Unit SizeEach