NIMBUS™ Target Preparation Instrument
Applied Biosystems™
NIMBUS™ Target Preparation Instrument
We have partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUSRead more
Have Questions?
Catalog NumberQuantity
00-0401Ea.
Catalog number 00-0401
Price (MXN)
-
Quantity:
Ea.
We have partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUS Target Preparation Instrument is industry-recognized for its high-end manufacturing and superior pipetting performance at an affordable price.
Axiom™ 96-sample automated target preparation
We have automated the target preparation steps for the Axiom 2.0 assay, including DNA amplification, fragmentation, purification, resuspension, hybridization preparation, and stain preparation for the GeneTitan™ MC Instrument.
The Axiom target preparation on the automated liquid-handling workstation helps minimize run-to-run variability and the labor burden associated with complex manual pipetting, helping to improve test reproducibility and laboratory efficiency.
Target preparation for Axiom Genotyping Solution is now effortless with Axiom 96-sample automated target preparation, which offers:
• Small footprint, sleek design, and industry-recognized engineering
• Advanced pipetting technology for precise yet flexible liquid handling
• Workflow designed to accommodate 96 samples
• Laptop with easy-to-use software interface including visual cues for each assay step
Pipetting technology
The NIMBUS Target Preparation Instrument represents the latest in compact, high-speed automated multi-channel pipetting. It features a 1 mL 96-channel multi-pipetting head for plate-based pipetting using a combination of capacitance-based liquid level detection and pressure-based liquid level detection for unparalleled performance.
Learn more about advanced technology and engineering of NIMBUS.Custom-designed robotics
The NIMBUS Target Preparation Instrument, co-developed with Hamilton Robotics, features an automation deck optimized for use with the Axiom assay protocol and an automated workflow that is compatible with Axiom 2.0 Reagent Kits. The on-deck thermoshake, cooling block, and customized trash chute provide convenience and support for assay robustness. The compact size allows the flexibility to place the instrument on virtually any benchtop.
Software
The NIMBUS Target Preparation Instrument is provided with a Dell™ laptop computer that is powered by an easy-to-use interface. The high-resolution visual display and simple on-screen instructions provide simple visual cues throughout assay progression.
Related Links
CytoScan™ Automation Starter Kit for NIMBUS™
CytoScan™ Consumables Kit for NIMBUS™
Axiom™ Starter Kit for NIMBUS
Axiom™ Consumables Kit for NIMBUS
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions20.0 in. (50.8 cm) x 37.4 in. (94.9 cm) x 30.0 in. maximum (76.2 cm)
For Use With (Application)Post-PCR automated target preparation
QuantityEa.
Weight (English)130 lb. (58.9 kg)
Product TypePost-PCR Target Preparation Instrument
Unit SizeEach