aminoxyTMTsixplex™ Label Reagent Set
Thermo Scientific™

aminoxyTMTsixplex™ Label Reagent Set

The carbonyl-reactive Thermo Scientific™ aminoxyTMT™ (Tandem Mass Tag™) Label Reagents enable multiplexed characterization and quantitation of carbonyl-containing biomolecules (carbohydrates, steroids, oxidized proteins) by mass spectrometry (MS).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
904025 x 0.2 mg (per tag)
904011 x 0.2 mg (per tag)
Catalog number 90402
Price (MXN)
-
Quantity:
5 x 0.2 mg (per tag)
Request bulk or custom format

The aminoxyTMTsixplex Label Reagents share an identical structure but contain different numbers of 13C and 15N isotopes in the mass reporter. The optimized labeling procedure can be completed in one hour. After tag labeling, samples are quenched and cleaned-up using HILIC solid-phase extraction before mass spectrometry analysis. The six compounds of the aminoxyTMTsixplex™ Reagent Set have the same mass (i.e., isobaric) and chemical structure (carbonyl-reactive aminoxy group, spacer arm, and mass reporter). However, the specific distribution of 13C and 15N isotopes on either side of the HCD or ETD MS/MS fragmentation site in each reagent results in a unique reporter mass (126-131Da) in the low mass region.

This set of reporter ions is used to measure the relative abundance of labeled molecules in a combined (multiplexed) MS sample representing six different treatment conditions. For glycobiology MS applications, the reagents enable quantitative profiling of glycan isoforms and discovery of glycan biomarkers; they provide improved ionization of labeled glycans for increased sensitivity and better retention of labeled glycans by reversed-phase liquid chromatography (RPLC). The aminoxyTMT reagents may be used to quantify a broad range of biologically important molecules including carbohydrates, steroids, or oxidized proteins.

Features:

  • Quantitative – enables relative quantitation of glycans or other carbonyl-containing biomolecules from multiple samples derived from cells, tissues or biological fluids
  • Stable – oxime bond and products formed by labeling reaction are stable and don't require an additional reduction step
  • Efficient – achieve labeling efficiency greater than 90% in one hour
  • Sensitive – 20-fold increase in signal-to-noise compared to unlabeled native glycan MS analysis
  • Multiplex – able to identify and characterize up to six samples concurrently
  • Optimized – procedure and reagents optimized for excellent labeling efficiency and recovery of glycans

Includes:

  • Label reagent sets contain sufficient aminoxyTMT tags for five sixplex experiments

Requires:

  • PNGase F (or PNGase A) glycosidase and Waters Oasis™ HLB columns

Applications:

  • Multiplex up to 6 different samples concurrently in a single experiment
  • Relative quantitation of glycans
  • Study of structural diversity of protein glycosylation
  • Study of glycosylation in cell signaling and regulation
  • Study of cancer progression, biomarker discovery and analysis of biotherapeutics

Native glycans are difficult to study by mass spectrometry because of their poor ionization efficiency. Quantitation of glycans is particularly challenging due to the lack of standards for all naturally occurring glycans and difficulties reproducibly quantifying multiple samples. The aminoxy group has better reactivity with carbonyls and better stability of the labeled product compared to hydrazides. Labeling with the aminoxyTMT reagents improves ionization of glycans, thus improving sensitivity, and enables relative quantitation of glycans for up to six samples concurrently.

These products are subject to a limited use label license.

For Research Use Only. Not for use in diagnostic procedures.

The Tandem Mass Tag (TMT) Reagent family consists of TMTzero, TMTduplex, TMTsixplex and TMT10plex sets which are specially designed to enable a rapid and cost-effective transition from method development to high-throughput protein quantitation. The TMTzero tag allows testing and optimization of sample preparation, labeling, fractionation and MS fragmentation for peptide identification and reporter detection without using the more costly isotope-labeled compounds. The TMTsixplex reagent set allows sixplex protein profiling for multiple conditions, including time courses, dose responses, replicates or multiple sample comparisons. Each TMT tag is based on the same chemical structure, eliminating the need to modify labeling conditions or HPLC separation conditions between experiments.

References:

  1. Atwood III, J.A., et al. (2008). Quantitation by Isobaric Labeling: Applications to Glycomics. J Proteome Res 7(1), 367–374.
  2. Prien JM, et al. (2010). Mass spectrometric-based stable isotopic 2-aminobenzoic acid glycan mapping for rapid glycan screening of biotherapeutics. Anal Chem. 82(4) 1498-508.
  3. Snovida et al., Applications of Aldehyde-Reactive Thermo Scientific Tandem Mass Tag (TMT) Reagents for Mass Spectrometry-based Quantitative Glycomics. ASMS 2013 conference proceedings.
Specifications
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeTandem Mass Tag™ (TMT) Labels
Quantity5 x 0.2 mg (per tag)
Workflow StepProtein Labeling
Detection MethodMass Spectrometry
Product LineaminoxyTMTsixplex
Product TypeaminoxyTMTsixplex™ Label Reagent Set
Starting MaterialCell Lysate
Unit SizeEach
Contents & Storage
Sufficient for: Five sixplex (5 x 6-way) experiments
• aminoxyTMT6-126 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-127 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-128 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-129 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-130 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-131 Label Reagent, 5 x 0.2 mg

Store at –20°C.