Alexa Fluor&trade; 488 C<sub>5</sub> Maleimide
Alexa Fluor&trade; 488 C<sub>5</sub> Maleimide
Alexa Fluor&trade; 488 C<sub>5</sub> Maleimide
Alexa Fluor&trade; 488 C<sub>5</sub> Maleimide
Invitrogen™

Alexa Fluor™ 488 C5 Maleimide

Alexa Fluor™ 488 is a bright, green fluorescent dye with excitation ideally suited to the 488 nm laser line. UsedRead more
Have Questions?
Catalog NumberQuantity
A102541 mg
Catalog number A10254
Price (USD)
464.00
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
464.00
Each
Alexa Fluor™ 488 is a bright, green fluorescent dye with excitation ideally suited to the 488 nm laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 488 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 488 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The maleimide derivative of Alexa Fluor™ 488 is the most popular tool for conjugating the dye to a thiol group on a protein, oligonucleotide thiophosphate, or low molecular weight ligand. The resulting Alexa Fluor™ 488 conjugates exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ maleimide:

Fluorophore label: Alexa Fluor™ 488 dye
Reactive group: maleimide
Reactivity: thiol groups on proteins and ligands, oligonucleotide thiophosphates
Ex/Em of the conjugate: 493/516 nm
Extinction coefficient: 72,000 cm-1M-1
Spectrally similar dyes: Fluorescein (FITC), Cy™2
Molecular weight: 720.66

Typical Conjugation Reaction
The protein should be dissolved at a concentration of 50-100 μM in a suitable buffer (10-100 mM phosphate, Tris, or HEPES) at pH 7.0-7.5. In this pH range, the protein thiol groups are sufficiently nucleophilic that they react almost exclusively with the reagent in the presence of the more numerous protein amine groups, which are protonated and relatively unreactive. We recommend reducing any disulfide bonds at this point using a 10-fold molar excess of reducing agent such as DTT or TCEP. Excess DTT must be removed by dialysis and subsequent thiol-modification should be carried out under oxygen-free conditions to prevent reformation of the disulfide bonds; these precautions are not necessary when using TCEP prior to maleimide conjugation.

The Alexa Fluor™ maleimide is typically dissolved in high-quality anhydrous dimethylsulfoxide (DMSO) at a concentration of 1-10 mM immediately prior to use, and stock solutions should be protected from light as much as possible. Generally, this stock solution is added to the protein solution dropwise while stirring to produce approximately 10-20 moles of reagent per mole of protein, and the reaction is allowed to proceed at room temperature for 2 hours or at 4°C overnight, protected from light. Any unreacted thiol-reactive reagent can be consumed by adding excess glutathione, mercaptoethanol, or other soluble low molecular weight thiol.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
Emission516 nm
Excitation493 nm
Label or DyeAlexa Fluor™ 488
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

What are the signal intensity differences between Alexa Fluor 350 dye and Alexa Fluor 488 dye?

In general, blue fluorescent dyes are not as bright as other dyes further along the color spectrum. Blue dyes are structurally smaller and have lower extinction coefficients, so they are typically not as bright compared to the green, red, and far red dyes.
When using an Alexa Fluor 350 secondary antibody, we recommend that you use it for highly expressed targets and at a higher concentration than what is typically required for green or red secondary antibodies.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (100)

Citations & References
Abstract
Authors:
Journal:
PubMed ID:11114349
Distance measurements within a concatamer of the plasma membrane Cl?/HCO3? exchanger, AE1.
Authors:Basu A, Mazor S, Casey JR,
Journal:Biochemistry
PubMed ID:20828148
'AE1, which exists in the erythrocyte plasma membrane as a noncovalent dimer, facilitates transmembrane Cl?/HCO3? exchange. Here a concatamer of AE1 (two AE1 monomers fused via a two-residue linker to form an intramolecular dimer) was designed to facilitate fluorescence resonance energy transfer (FRET) studies. The concatameric protein (AE1·AE1) was expressed ... More
Interaction of arginine-rich peptides with membrane-associated proteoglycans is crucial for induction of actin organization and macropinocytosis.
Authors:Nakase I, Tadokoro A, Kawabata N, Takeuchi T, Katoh H, Hiramoto K, Negishi M, Nomizu M, Sugiura Y, Futaki S
Journal:Biochemistry
PubMed ID:17209559
'Arginine-rich peptides, including octaarginine (R8), HIV-1 Tat, and branched-chain arginine-rich peptides, belong to one of the major classes of cell-permeable peptides which deliver various proteins and macromolecules to cells. The importance of the endocytic pathways has recently been demonstrated in the cellular uptake of these peptides. We have previously shown ... More
Exocytotic insertion of calcium channels constrains compensatory endocytosis to sites of exocytosis.
Authors:Smith RM, Baibakov B, Ikebuchi Y, White BH, Lambert NA, Kaczmarek LK, Vogel SS
Journal:J Cell Biol
PubMed ID:10684256
'Proteins inserted into the cell surface by exocytosis are thought to be retrieved by compensatory endocytosis, suggesting that retrieval requires granule proteins. In sea urchin eggs, calcium influx through P-type calcium channels is required for retrieval, and the large size of sea urchin secretory granules permits the direct observation of ... More
Actin binding to the central domain of WASP/Scar proteins plays a critical role in the activation of the Arp2/3 complex.
Authors:Kelly AE, Kranitz H, Dötsch V, Mullins RD
Journal:J Biol Chem
PubMed ID:16403731
'The Arp2/3 complex nucleates and cross-links actin filaments at the leading edge of motile cells, and its activity is stimulated by C-terminal regions of WASP/Scar proteins, called VCA domains. VCA domains contain a verprolin homology sequence (V) that binds monomeric actin and central (C) and acidic sequences (A) that bind ... More