Alexa Fluor&trade; 546 C<sub>5</sub> Maleimide
Invitrogen™

Alexa Fluor™ 546 C5 Maleimide

Alexa Fluor™ 546 is a bright, orange fluorescent dye that can be excited using the 488 nm or 532 nmRead more
Have Questions?
Catalog NumberQuantity
A102581 mg
Catalog number A10258
Price (USD)
484.00
Each
Quantity:
1 mg
Price (USD)
484.00
Each
Alexa Fluor™ 546 is a bright, orange fluorescent dye that can be excited using the 488 nm or 532 nm laser lines. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 546 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 546 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The maleimide derivative of Alexa Fluor™ 546 is the most popular tool for conjugating the dye to a thiol group on a protein, oligonucleotide thiophosphate, or low molecular weight ligand. The resulting Alexa Fluor™ 546 conjugates exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ maleimide:

Fluorophore label: Alexa Fluor™ 546 dye
Reactive group: maleimide
Reactivity: thiol groups on proteins and ligands, oligonucleotide thiophosphates
Ex/Em of the conjugate: 554/570 nm
Extinction coefficient: 93,000 cm-1M-1
Spectrally similar dyes: Rhodamine red, Cy3
Molecular weight: 1034.37

Typical Conjugation Reaction
The protein should be dissolved at a concentration of 50-100 μM in a suitable buffer (10-100 mM phosphate, Tris, or HEPES) at pH 7.0-7.5. In this pH range, the protein thiol groups are sufficiently nucleophilic that they react almost exclusively with the reagent in the presence of the more numerous protein amine groups, which are protonated and relatively unreactive. We recommend reducing any disulfide bonds at this point using a 10-fold molar excess of reducing agent such as DTT or TCEP. Excess DTT must be removed by dialysis and subsequent thiol-modification should be carried out under oxygen-free conditions to prevent reformation of the disulfide bonds; these precautions are not necessary when using TCEP prior to maleimide conjugation.

The Alexa Fluor™ maleimide is typically dissolved in high-quality anhydrous dimethylsulfoxide (DMSO) at a concentration of 1-10 mM immediately prior to use, and stock solutions should be protected from light as much as possible. Generally, this stock solution is added to the protein solution dropwise while stirring to produce approximately 10-20 moles of reagent per mole of protein, and the reaction is allowed to proceed at room temperature for 2 hours or at 4°C overnight, protected from light. Any unreacted thiol-reactive reagent can be consumed by adding excess glutathione, mercaptoethanol, or other soluble low molecular weight thiol.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
Emission570 nm
Excitation554 nm
Label or DyeAlexa Fluor™ 546
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (17)

Citations & References
Abstract
Cofilin promotes stimulus-induced lamellipodium formation by generating an abundant supply of actin monomers.
Authors:Kiuchi T, Ohashi K, Kurita S, Mizuno K
Journal:J Cell Biol
PubMed ID:17470633
'Cofilin stimulates actin filament disassembly and accelerates actin filament turnover. Cofilin is also involved in stimulus-induced actin filament assembly during lamellipodium formation. However, it is not clear whether this occurs by replenishing the actin monomer pool, through filament disassembly, or by creating free barbed ends, through its severing activity. Using ... More
The citrate carrier CitS probed by single-molecule fluorescence spectroscopy.
Authors:Kästner CN, Prummer M, Sick B, Renn A, Wild UP, Dimroth P
Journal:Biophys J
PubMed ID:12609868
'A prominent region of the Na(+)-dependent citrate carrier (CitS) from Klebsiella pneumoniae is the highly conserved loop X-XI, which contains a putative citrate binding site. To monitor potential conformational changes within this region by single-molecule fluorescence spectroscopy, the target cysteines C398 and C414 of the single-Cys mutants (CitS-sC398, CitS-sC414) were ... More
Pointed-end capping by tropomodulin3 negatively regulates endothelial cell motility.
Authors:Fischer RS, Fritz-Six KL, Fowler VM
Journal:J Cell Biol
PubMed ID:12707310
'Actin filament pointed-end dynamics are thought to play a critical role in cell motility, yet regulation of this process remains poorly understood. We describe here a previously uncharacterized tropomodulin (Tmod) isoform, Tmod3, which is widely expressed in human tissues and is present in human microvascular endothelial cells (HMEC-1). Tmod3 is ... More
Protein translocation through the anthrax toxin transmembrane pore is driven by a proton gradient.
Authors:Krantz BA, Finkelstein A, Collier RJ
Journal:J Mol Biol
PubMed ID:16343527
'Protective antigen (PA) from anthrax toxin assembles into a homoheptamer on cell surfaces and forms complexes with the enzymatic components: lethal factor (LF) and edema factor (EF). Endocytic vesicles containing these complexes are acidified, causing the heptamer to transform into a transmembrane pore that chaperones the passage of unfolded LF ... More
Labeling of DNA via rearrangement of S-2-aminoethyl phosphorothioates to N-(2-mercaptoethyl)phosphoramidates.
Authors:Chen M, Gothelf KV,
Journal:Org Biomol Chem
PubMed ID:18292883
The reaction of phosphorothioates in DNA with 2-bromoethylammonium bromide results in S-2-aminoethyl phosphorothioates, which can rearrange to N-(2-mercaptoethyl)phosphoramidates providing a facile method for the generation of site-specific thiol labeling of DNA sequences. The applicability of this method was demonstrated by conjugation of the thiolated DNA sequence with Na-(3-maleimidylpropionyl) biocytin and ... More