Are the vials of ArC Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD Fixable dead cell stain kits) (Cat. No. A10346, A10648) compatible with Vaporized Hydrogen Peroxide (VHP) sterilization?
While the polypropylene cap is compatible with Vaporized Hydrogen Peroxide (VHP) sterilization, we have not tested the compatibility of the Low-Density Polyethylene (LDPE) / Linear Low-Density Polyethylene (LLDPE) vials with VHP sterilization.