Alexa Fluor™ 594 Hydrazide, for microinjection
Invitrogen™

Alexa Fluor™ 594 Hydrazide, for microinjection

Alexa Fluor™ 594 Hydrazide is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketonesRead more
Have Questions?
Catalog NumberQuantity
A10442
also known as A-10442
125 μL
Catalog number A10442
also known as A-10442
Price (USD)
442.00
Each
Quantity:
125 μL
Price (USD)
442.00
Each
Alexa Fluor™ 594 Hydrazide is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins. This version is formatted as a ready-to-use solution that is dissolved in a 200 mM KCl solution and filter sterilized.

Alexa Fluor™ 594 is a bright, red fluorescent dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 594 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 594 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

Detailed information about this AlexaFluor™ hydrazide:

• Fluorophore label : Alexa Fluor™ 594 dye
• Reactive group: hydrazide
• Reactivity: Aldehydes or keytones in polysaccharides or glycoproteins
• Ex/Em of the conjugate: 588/613 nm
• Extinction coefficient: 97,000 cm-1M-1
• Spectrally similar dyes: Texas Red
• Molecular weight: 758.79

Cell Tracking and Tracing Applications
Alexa Fluor™ hydrazides and hydroxlamines are useful as low molecular weight, membrane-impermeant, aldehyde-fixable cell tracers, exhibiting brighter fluorescence and greater photostability than cell tracers derived from other spectrally similar fluorophores. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent. Learn more about cell tracking and tracing.

Glycoprotein and Polysaccharide Labeling Applications
The Alexa Fluor™ hydrazides and hydroxlamines are reactive molecules that can be used to add a fluorescent label to biomolecules containing aldehydes or ketones. Aldehydes and ketones can be introduced into polysaccharides and glycoproteins by periodate-mediated oxidation of vicinal diols. Galactose oxidase can also be used to oxidize terminal galactose residues of glycoproteins to aldehydes.

Hydrazide vs Hydroxylamine
Hydrazine derivatives react with ketones and aldehydes to yield relatively stable hydrazones. Hydroxylamine derivatives (aminooxy compounds) react with aldehydes and ketones to yield oximes. Oximes are superior to hydrazones with respect to hydrolytic stability. Both hydrazones and oximes can be reduced with sodium borohydride (NaBH4) to further increase the stability of the linkage.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityCarboxylic Acid, Ketone, Aldehyde
Emission613 nm
Excitation588 nm
Label or DyeAlexa Fluor™ 594
Product TypeHydrazide
Quantity125 μL
Reactive MoietyAmine, Hydrazide
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

What dye can I use that is non-reactive and can show an injection site?

The non-reactive Alexa Fluor and Alexa Fluor hydrazide derivatives may be used for injection site visualization. Other options include the fluorescent polystyrene microspheres, FluoSpheres, and dye-conjugated dextrans. The hydrazide derivatives and 'fixable' dextrans are retained by cross-linking using an aldehyde-based fixative.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (61)

Citations & References
Abstract
Supersensitive Ras activation in dendrites and spines revealed by two-photon fluorescence lifetime imaging.
Authors:Yasuda R, Harvey CD, Zhong H, Sobczyk A, van Aelst L, Svoboda K
Journal:Nat Neurosci
PubMed ID:16429133
'To understand the biochemical signals regulated by neural activity, it is necessary to measure protein-protein interactions and enzymatic activity in neuronal microcompartments such as axons, dendrites and their spines. We combined two-photon excitation laser scanning with fluorescence lifetime imaging to measure fluorescence resonance energy transfer at high resolutions in brain ... More
Alexa dyes, a series of new fluorescent dyes that yield exceptionally bright, photostable conjugates.
Authors:Panchuk-Voloshina N, Haugland RP, Bishop-Stewart J, Bhalgat MK, Millard PJ, Mao F, Leung WY, Haugland RP
Journal:J Histochem Cytochem
PubMed ID:10449539
'Alexa 350, Alexa 430, Alexa 488, Alexa 532, Alexa 546, Alexa 568, and Alexa 594 dyes are a new series of fluorescent dyes with emission/excitation spectra similar to those of AMCA, Lucifer Yellow, fluorescein, rhodamine 6G, tetramethylrhodamine or Cy3, lissamine rhodamine B, and Texas Red, respectively (the numbers in the ... More
Rapid and reversible chemical inactivation of synaptic transmission in genetically targeted neurons.
Authors:Karpova AY, Tervo DG, Gray NW, Svoboda K
Journal:Neuron
PubMed ID:16337911
'Inducible and reversible silencing of selected neurons in vivo is critical to understanding the structure and dynamics of brain circuits. We have developed Molecules for Inactivation of Synaptic Transmission (MISTs) that can be genetically targeted to allow the reversible inactivation of neurotransmitter release. MISTs consist of modified presynaptic proteins that ... More
The kinetics of phagosome maturation as a function of phagosome/lysosome fusion and acquisition of hydrolytic activity.
Authors:Yates RM, Hermetter A, Russell DG
Journal:Traffic
PubMed ID:15813751
'Professional phagocytes function at the hinge of innate and acquired immune responses by internalizing particulate material that is digested and sampled within the phagosome of the cell. Despite intense interest, assays to measure phagosome maturation remain insensitive and few in number. In this current study, we describe three novel assays ... More
Intracellular fluid flow in rapidly moving cells.
Authors:Keren K, Yam PT, Kinkhabwala A, Mogilner A, Theriot JA,
Journal:Nat Cell Biol
PubMed ID:19767741
'Cytosolic fluid dynamics have been implicated in cell motility because of the hydrodynamic forces they induce and because of their influence on transport of components of the actin machinery to the leading edge. To investigate the existence and the direction of fluid flow in rapidly moving cells, we introduced inert ... More