StemPro™ Neural Supplement - FAQs
StemPro™ Neural Supplement - FAQs
View additional product information for StemPro™ Neural Supplement - FAQs (A1050801)
5 product FAQs found
Which cells can I use StemPro Neural Supplement with?
StemPro Neural Supplement can be used with human and rat NSCs, GPCs, and some neuronal progenitors.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the key applications of StemPro Neural Supplement?
StemPro Neural Supplement supports the growth and expansion of neural stem cells and progenitor cells. It also supports the survival of neurons and differentiation of progenitor cells to astrocytes, oligodendrocytes, and neurons. Besides, its application can also be extended to transplantation studies and screening of drug candidates.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do I need to add other supplements when using StemPro Neural Supplement?
Mixing of StemPro Neural Supplement with a basal medium such as Knockout DMEM/F-12 or Neurobasal medium and the addition of appropriate quantities of recommended growth factors yields a complete medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the intended uses of StemPro Neural Supplement?
StemPro Neural Supplement is an optimized serum-free additive for the growth and expansion of neural stem and progenitor cells. The intended use is "For research use only".
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is StemPro Neural Supplement serum-free?
Yes, StemPro Neural Supplement is serum-free.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.