StemPro™ Neural Supplement
Gibco™

StemPro™ Neural Supplement

StemPro Neural Supplement is an optimal, convenient, and cost-effective serum-free supplement for the proliferation of mammalian neural stem and progenitorRead more
Have Questions?
Catalog NumberQuantity
A105080110 mL
Catalog number A1050801
Price (TWD)
8,610.00
Online offer
Ends: 31-Dec-2026
12,300.00
Save 3,690.00 (30%)
Each
Quantity:
10 mL
Price (TWD)
8,610.00
Online offer
Ends: 31-Dec-2026
12,300.00
Save 3,690.00 (30%)
Each
StemPro Neural Supplement is an optimal, convenient, and cost-effective serum-free supplement for the proliferation of mammalian neural stem and progenitor cells such as human and rat neural stem cells (NSC) and glial progenitor cells (GPC). Due to their scarce availability, the in vitro expansion of these cells has been a challenge to academic and clinical researchers. When combined with the appropriate basal medium and growth factors, StemPro Neural Supplement not only supports the expansion of neuronal progenitor cells but also enables the cells to retain their progenitor status and potential to differentiate into neurons and glial cells.

In recent years, there has been an increased scientific interest in studying neural progenitor cells due to their important role in neurogenesis. Recent advances have also pointed to the potential use of neural progenitor cells as effective therapies for neurodegenerative diseases, such as dopaminergic neural progenitor cells for Parkinson’s Disease and oligoprogenitor cells for amyotrophic lateral sclerosis (ALS) and multiple sclerosis (MS).

For established protocols using StemPro Neural Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.
Specifications
Cell TypeNeural Stem Cells
Concentration50 X
Culture TypeMammalian Cell Culture
Endotoxin Level≤ 50 EU/ml
For Use With (Application)Academic Research, Clinical Research
Product LineStemPro
Quantity10 mL
Serum LevelSerum-Free
Shelf Life12 Months
Shipping ConditionDry Ice
SourceSerum-Free
Tested ForEndotoxin, Performance, Sterility, pH
ClassificationSerum-free
FormLiquid
Product TypeNeural Supplement
SterilitySterile
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.

Frequently asked questions (FAQs)

Which cells can I use StemPro Neural Supplement with?

StemPro Neural Supplement can be used with human and rat NSCs, GPCs, and some neuronal progenitors.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What are the key applications of StemPro Neural Supplement?

StemPro Neural Supplement supports the growth and expansion of neural stem cells and progenitor cells. It also supports the survival of neurons and differentiation of progenitor cells to astrocytes, oligodendrocytes, and neurons. Besides, its application can also be extended to transplantation studies and screening of drug candidates.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I need to add other supplements when using StemPro Neural Supplement?

Mixing of StemPro Neural Supplement with a basal medium such as Knockout DMEM/F-12 or Neurobasal medium and the addition of appropriate quantities of recommended growth factors yields a complete medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What are the intended uses of StemPro Neural Supplement?

StemPro Neural Supplement is an optimized serum-free additive for the growth and expansion of neural stem and progenitor cells. The intended use is "For research use only".

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is StemPro Neural Supplement serum-free?

Yes, StemPro Neural Supplement is serum-free.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.