Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Secondary Antibodies ›
          • Goat anti-Chicken IgY Secondary Antibodies

          Invitrogen

          Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor™ 488

          97 Published Figures
          View all (39) Goat anti-Chicken IgY antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor™ 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (97)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Chicken IgY (H+L) Secondary Antibody (A-11039) in ICC/IF

          Mouse intestine cryosection showing basement membranes labeled with our chicken IgY anti-fibronectin antibody (Product # A21316) and the Alexa Fluor® 488 Goat Anti-Chicken IgG (Product # A-11039, green). Goblet cells and crypt cells were labeled with Alexa Fluor® 594 wheat germ agglutinin (Product # W11262, red). The microvillar brush border and smooth muscle layers were visualized with Alexa Fluor® 680 pha... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunocytochemistry (ICC/IF)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)
          Chicken IgY (H+L) Secondary Antibody in Immunohistochemistry (IHC)

          Product Details

          A-11039

          Applications
          Tested Dilution

          Immunohistochemistry (IHC)

          1-10 µg/mL

          Immunocytochemistry (ICC/IF)

          1-10 µg/mL

          Flow Cytometry (Flow)

          1-10 µg/mL
          Product Specifications

          Species Reactivity

          Chicken

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Secondary Antibody

          Immunogen

          Gamma Immunoglobins Heavy and Light chains

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Alexa Fluor 546 
          • Alexa Fluor 555 
          • Alexa Fluor 568 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • AP
          • Biotin 
          • FITC
          • HRP
          • Texas Red
          • TRITC
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          1 mg

          Purification

          purified

          Storage buffer

          PBS, pH 7.5

          Contains

          5mM sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2534096

          Target

          IgY

          Antibody Form

          Whole Antibody

          Product Specific Information

          Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 488 dye is a bright, green-fluorescent dye with excitation ideally suited to the 488 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 488 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 488 dye molecules can be attached to proteins at high molar ratios without significant self-quenching, enabling brighter conjugates and more sensitive detection. The degree of labeling for each conjugate is typically 2-8 fluorophore molecules per IgG molecule; the exact degree of labeling is indicated on the certificate of analysis for each product lot.

          Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. For the fluorophore-labeled antibodies a final concentration of 1-10 µg/mL should be satisfactory for most immunohistochemistry and flow cytometry applications.

          Product will be shipped at Room Temperature.

          Target Information

          Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody A-11039 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline