CD FortiCHO™ Medium
Gibco™

CD FortiCHO™ Medium

CD FortiCHO™ medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHORead more
Have Questions?
Catalog NumberQuantity
A11483011 L
Catalog number A1148301
Price (USD)
188.00
Each
Quantity:
1 L
Price (USD)
188.00
Each
CD FortiCHO™ medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment. The protein-free, animal origin component-free formulation provides consistency and helps reduce the need to screen for adventitious agents. With CD FortiCHO™ medium cell cultures last 2+ days longer than competitive alternatives, which means decreased HCP (Host Cell Protein) concentration for removal in downstream purification.

CD FortiCHO™ medium is fortified to support maximal recombinant CHO cell growth and protein expression in suspension batch culture, offering 120% greater growth and production than select competitive media with simple glucose supplementation. CD FortiCHO™ medium can provide up to 75% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with CD EfficientFeed™ C nutrient supplement.

When to Use CD FortiCHO™ Medium
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

When to Consider Another Gibco™ Medium

Consider CD OptiCHO™ medium when:
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky,” i.e. hard to grow or adapt

Consider CD CHO medium when:
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

Consider CD DG44 when:
• You use parental DG44 or DXB11 CHO cells (dhfr-)

Dry Media Formulations
Dynamis AGT Medium is the comparable dry format for scale up.

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineCHO
Concentration1 X
Endotoxin LevelLow
FormatBottle
Included AntibioticsNo antibiotics
Product LineGibco
Product TypeCD OptiCHO AGT Medium
Quantity1 L
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
With AdditivesSodium Pyruvate, Sodium Bicarbonate
Without AdditivesNo Glutamine, No Phenol Red
Unit SizeEach
Contents & Storage
Storage:
Store in the dark at +2 to +8°C. Guaranteed stable for 12 months when properly stored.