Search
Search

| Catalog Number | Quantity |
|---|---|
| A2617501 | 10 L |
| A2617503 | 10 kg |
| A2617504 | 1 L |
| A2617502 | 100 L |
Gibco Dynamis AGT dry format medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment. The chemically defined, protein-free, animal origin component-free formulation provides the power to achieve high titers, start process development faster, and streamline or simplify transfer to manufacturing scale.
Power to achieve higher titers
Dynamis AGT Medium is fortified to help maximize recombinant CHO cell growth and protein expression in suspension batch culture, offering 170% greater production than select competitive media with simple glucose supplementation by day 14. Cultures grown in Dynamis AGT Medium extend to even higher titers demonstrating good cell viability out to day 21. This medium can also provide up to 60% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with EfficientFeed C+ Supplement (Cat. No. A2503104).
Power to start process development faster
As a fortified medium, Dynamis AGT provides the power to get to a high cell density over 10 million cells per mL faster than competitive products and sustains that cell density longer by just maintaining glucose levels. The viable cell growth achieved make it a good choice for scale-up if you have started with CD FortiCHO medium. When paired with EfficientFeed C+ Supplement (A2503104), it provides a fast and efficient way to develop a process.
The power to streamline transfer to manufacturing
By choosing Dynamis AGT Medium, you obtain not only the benefits of the medium formulation, but also the benefits of AGT dry format technology. This all-in-one product is designed to simplify scale-up and manufacturing by being easy to reconstitute (just add water), with no adjustments of pH or osmolarity necessary. This can help provide efficiencies through supply chain simplicity, reconstitution simplicity, and therefore help reduce expense.
Use Dynamis AGT Medium when:
When to consider another Gibco medium
Consider CD OptiCHO Medium (12681011) when:
Consider CD CHO Medium (10743029) when:
Consider CD DG44 Medium (12610010) when:
Design your own media specific to your needs
Whether it's a bench-level experiment or large-scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco cGMP Media Custom Services, you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Gibco is a registered trademark of ThermoFisher Scientific Inc. Dynamis and CD FortiCHO are trademarks of Thermo Fisher Scientific Inc., ActiCHO is a trademark of Cytiva, BalanCD is a registered trademark of Irvine Scientific Sales Co., Cellvento is a registered trademark of Merck KGAA, EXCELL is a registered trademark of Millipore Sigma, HyCell is a registered trademark of Cytiva, and PowerCHO is a registered trademark of Lonza.