Alkaline Phosphatase Live Stain is a stem cell imaging product that allows users to differentially stain pluripotent stem cells (PSCs).Read more
Have Questions?
Catalog Number
Quantity
A14353
50 μl
Catalog number A14353
Price (TWD)
13,000.00
Online offer
Ends: 30-Sep-2026
17,000.00
Save 4,000.00 (24%)
Each
Quantity:
50 μl
Price (TWD)
13,000.00
Online offer
Ends: 30-Sep-2026
17,000.00
Save 4,000.00 (24%)
Each
Alkaline Phosphatase Live Stain is a stem cell imaging product that allows users to differentially stain pluripotent stem cells (PSCs). The dye is a cell-permeable fluorescent substrate for alkaline phosphatase (AP) that is non-toxic to cells, diffusing out over the course of two hours. The Alkaline Phosphatase Live Stain consists of 50 μL of a 500X fluorescein-based dye in DMSO, sufficient for staining two 24-well plates, four 6 cm dishes or two 10 cm dishes.
This Alkaline Phosphatase Live Stain allows you to:
• Identify pluripotent stem cells with a strong signal above background • Stain PSCs typically in 20 minutes or less • Use the first alkaline phosphatase stain marketed that maintains cell viability
Alkaline Phosphatase Pluripotent Stem Cell Detection Method AP is a phenotypic marker of PSCs, including undifferentiated embryonic stem cells (ESCs), induced pluripotent stem cells (iPSCs), and embryonic germ cells (EGCs). While AP is expressed in most cell types, its expression is highly elevated in PSCs. AP staining has therefore been used to differentially stain PSCs to easily distinguish them from mouse embryonic fibroblasts (MEFs) used as feeders and parental fibroblasts commonly used in reprogramming experiments.
Screen for Pluripotency Without Sacrificing Precious Colonies The Alkaline Phosphatase Live Stain is ideal for screening colonies during early stages of the reprogramming workflow since the stain maintains stem cell viability. It can also be used in later stages as a negative selection tool for identifying undifferentiated cells.
Stain Cells Quickly The Alkaline Phosphatase Live Stain utilizes an easy, non-permanent, cell-viable protocol for identifying PSCs in your experiments. Simply dilute the dye in media, apply to cells, gently wash, and the cells are ready for fluorescent imaging.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Use With (Equipment)Floid Cell Imaging System, Fluorescence Microscope
Product TypeAlkaline Phosphatase Live Stain
Unit SizeEach
Contents & Storage
Contains 1 amber vial. Store at -20°C.
Frequently asked questions (FAQs)
How can I prevent poor or dim staining when using Alkaline Phosphatase Live Stain?
Serum or serum replacement components in the growth medium may cause background and this can result in poor or dim staining. After the removal of the growth medium, gently wash the culture with pre-warmed DMEM/F-12 (Cat. No. 10565-018) for 5 minutes. Aspirate and repeat once before adding the AP Live Stain. To further decrease background staining, perform 3 separate washes of 5 minutes each with DMEM/F-12 for a total of 15 minutes following the staining, and visualize immediately.
How should Alkaline Phosphatase Live Stain be stored?
Store AP Live Stain at -20 degrees C in the freezer and thaw at room temperature. Avoid repeated freeze/thaw cycles and aliquot the AP Live Stain into smaller volumes if necessary. Always protect from light and avoid extended exposure at room temperature and atmospheric conditions.
Can I use Alkaline Phosphatase Live Stain in my reprogramming experiments?
AP Live Stain is ideal for screening colonies during early stages of reprogramming since it selectively stains PSCs while maintaining cell viability. It can be used in later stages of reprogramming to identify undifferentiated cells for the selection of iPSCs for further cultivation. AP Live Stain was developed specifically for use on live cultures for cell maintenance and is qualified to be free of mycoplasma and bioburden, and exhibits extremely low endotoxin levels.
How long should I wait before using Alkaline Phosphatase Live stain on my cells again?
Colonies may be restained as early as 24 hours after the initial staining. It is recommended that cells are allowed to recover properly to obtain consistent results.
Novel Live Alkaline Phosphatase Substrate for Identification of Pluripotent Stem Cells.
Authors:Singh U, Quintanilla RH, Grecian S, Gee KR, Rao MS, Lakshmipathy U,
Journal:Stem Cell Rev
PubMed ID:22426885
Characterization of pluripotent stem cells.
Authors:Martí M, Mulero L, Pardo C, Morera C, Carrió M, Laricchia-Robbio L, Esteban CR, Izpisua Belmonte JC,
Journal:Nat Protoc
PubMed ID:23306458
Characterization of pluripotent stem cells is required for the registration of stem cell lines and allows for an impartial and objective comparison of the results obtained when generating multiple lines. It is therefore crucial to establish specific, fast and reliable protocols to detect the hallmarks of pluripotency. Such protocols should ... More
Immune Defense Protein Expression in Highly Purified Mouse Lung Epithelial Cells.
Authors:Sinha M, Lowell CA
Journal:Am J Respir Cell Mol Biol
PubMed ID:26574781
Lung epithelial cells play critical roles in initiating and modulating immune responses during pulmonary infection or injury. To better understand the spectrum of immune response-related proteins present in lung epithelial cells, we developed an improved method of isolating highly pure primary murine alveolar type (AT) II cells and murine tracheal ... More