How can I prevent poor or dim staining when using Alkaline Phosphatase Live Stain?
Serum or serum replacement components in the growth medium may cause background and this can result in poor or dim staining. After the removal of the growth medium, gently wash the culture with pre-warmed DMEM/F-12 (Cat. No. 10565-018) for 5 minutes. Aspirate and repeat once before adding the AP Live Stain. To further decrease background staining, perform 3 separate washes of 5 minutes each with DMEM/F-12 for a total of 15 minutes following the staining, and visualize immediately.