RapidFinder™ Quant Equine Set
Thermo Scientific™
RapidFinder™ Quant Equine Set
The RapidFinder™ Quant Equine Set, when used with the RapidFinder™ Equine ID Kit, enables the fast and accurate quantification ofRead more
Have Questions?
Catalog number A15579
Price (USD)
-
The RapidFinder™ Quant Equine Set, when used with the RapidFinder™ Equine ID Kit, enables the fast and accurate quantification of horse meat in food and feed samples. Using the included standards, the kit enables quantification of as little as 0.05% horse meat DNA relative to other meat DNA in only a few hours.
• Simple—real-time PCR based assay, no electrophoresis needed
• Flexible—compatible with any food or feed sample, both raw and processed
• Reliable—includes equine DNA quantification standard
• Fast—from sample to result on the same day
How It Works
The kit enables the quantitative real-time PCR-based generation of a standard curve using the included Equus caballus DNA standard. Then assays for both equine DNA and all meat DNA, detected in the FAM™ channel, are run side by side on both food samples and the standards. This approach enables rapid and accurate quantification and reporting of the percentage of horse meat as compared to other meat species in a sample. Included in each reaction mix is an internal positive control detected in the VIC™ channel, which is used to verify the absence of PCR inhibitors.
The RapidFinder™ Quant Equine Set is designed to be used with the RapidFinder™ Equine ID Kit, and is optimized and validated for use with the Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument and the Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument.
Sample Preparation
DNA sample preparation for the RapidFinder™ Equine ID Kit and RapidFinder™ Quant Equine Set workflow is accomplished with the easy-to-use GMO Extraction Kit. Using 10 g of sample rather than smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.
Specifications
Product LineRapidFinder™
Quantity48 reactions
SpeciesEquine
No. of Reactions48 Reactions
Unit SizeEach
Contents & Storage
Animal Master Mix (store at -20°C)
General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use. Store protected from light)
Equine Standard (store at -20°C)