Kit de extracción de OMG
Thermo Scientific™
Kit de extracción de OMG
The GMO Extraction Kit enables fast and easy purification of DNA for use in food safety testing for genetically modifiedMás información
Have Questions?
Número de catálogoCantidad
446633650 preparaciones
Número de catálogo 4466336
Precio (MXN)
-
Cantidad:
50 preparaciones
The GMO Extraction Kit enables fast and easy purification of DNA for use in food safety testing for genetically modified organisms. The kit is capable of isolating highly purified DNA from a wide variety of different sample types without the use of toxic reagents. The benefits of the GMO Extraction Kit over other commercially-available products include:
• Performance—isolation of extremely pure DNA (A260/A280 >1.8)
• Specificity—adapted to food samples for transgenic material analysis
• Efficiency—total assay time is less than 2 hours, requiring minimal handling steps
The GMO Extraction Kit employs silica-based DNA purification using spin column technology to provide samples for GMO testing. The kit has been specifically adapted for transgenic material analysis of a wide array of sample matrices, including raw materials, processed food, and feed. Effectively removing inhibitors, the GMO Extraction Kit provides isolated, highly-purified DNA, quickly, reliably, and safely.
The development and manufacture of this kit occurs through a partnership with Health in Code, S.L., which has a long history of GMO research and development.
Other related products include:
Taqman™ Roundup Ready Soya Quantification Kit
TaqMan™ GMO Screening Kit
TaqMan™ GMO Maize Quantification Kit
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Tipo de producto finalADN genómico
Para utilizar con (aplicación)Real-Time Quantitative PCR (qPCR), Sequencing, PCR
Compatibilidad de alto rendimientoNo compatible con alto rendimiento (manual)
N.º de reacciones50 Isolations
Cantidad50 preparaciones
Tipo de muestraFood and Environmental, Fungi, Plant, Yeast
Cantidad de material de partida5-20 g
ObjetivoADN genómico
Tiempo de prueba2 hrs
Producción100 ng/μL
FormatoSpin Column
Isolation TechnologySilica Spin Column
Unit SizeEach