TaqMan™ Roundup Ready™ Soya Quantification Kit
Thermo Scientific™

TaqMan™ Roundup Ready™ Soya Quantification Kit

The TaqMan™ Roundup Ready™ Soya Quantification Kit is a food safety testing kit used to determine the percentage of RoundupRead more
Have Questions?
Catalog NumberQuantity
44663352 x 48 reactions
Catalog number 4466335
Price (USD)
-
Quantity:
2 x 48 reactions
The TaqMan™ Roundup Ready™ Soya Quantification Kit is a food safety testing kit used to determine the percentage of Roundup Ready™ soya against the total amount of soya present in a DNA sample. This real-time PCR kit contains all reagents (except for the template DNA) required for the quantification of Roundup Ready™ soya. The benefits of the TaqMan™ Roundup Ready™ Soya Quantification Kit over other commercially available products include:

• Efficiency—detection of small DNA fragments through use of Taqman™ MGB probes
• Sensitivity—quantification limit of 20 DNA copies and detection limit of 3 DNA copies each of Roundup Ready™ and soya DNA
• Flexibility—adapted to processed DNAs

The TaqMan™ Roundup Ready™ Soya Quantification Kit consists of plasmid-containing targets for the endogenous soy gene, known as lectin, and for the transgenic event (GTS-40-3-2) present in Roundup Ready™ soybean. This soybean accounts for approximately 80% of all soybean cultivated worldwide and is considered to be the most widespread transgenic event.

The TaqMan™ Roundup Ready™ Soya Quantification Kit is optimized and validated for use with  the Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument and the Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument.

The development and manufacture of this kit occurs through a partnership with Health in Code, S.L., which has a long history of GMO research and development.

Other related products include:
GMO Extraction Kit
TaqMan™ GMO Screening Kit
TaqMan™ GMO Maize Quantification Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Kit TypeRoundup Ready™ Soya Quantification Kit
No. of Reactions96 Reactions
Product LineTaqMan
Quantity2 x 48 reactions
Sample TypeFood
TargetLectin, GTS-40-3-2
Target SpecificityTarget-Specific
Detection MethodPrimer-probe
Unit SizeEach