TaqMan™ GMO Screening Kit
Thermo Scientific™
TaqMan™ GMO Screening Kit
The TaqMan™ GMO Screening Kit analyzes DNA extracted from food and feed samples for the presence of the main transgenicMás información
Have Questions?
Número de catálogoCantidad
44663344 x 48 reactions
Número de catálogo 4466334
Precio (MXN)
-
Cantidad:
4 x 48 reactions
The TaqMan™ GMO Screening Kit analyzes DNA extracted from food and feed samples for the presence of the main transgenic regulatory regions. This real-time PCR kit contains all reagents (except for the template DNA) required for the detection of genetically modified organisms (GMOs). In addition, the kit contains an Internal Positive Control (IPC) to identify possible PCR inhibition. The benefits of the Taqman™ GMO Screening Kit over other commercially available products include:
• Flexibility—GMOs detected in DNA obtained from ANY food or feed source
• Accuracy—an internal positive control (IPC) rules out inhibition during qPCR process
• Speed—results provided in less than 3 hours
The TaqMan™ GMO Screening Kit is capable of multiplex amplification of Endogenous Vegetal Control, Internal Positive Control, and these regulatory regions:
• P35S/CaMV
• TNOS/A. tumefaciens
• P34S/FMV
All of which make possible the detection of significant events such as soybean MON89788.
The TaqMan™ GMO Screening Kit provides all of the necessary reagents for testing 48 samples and is optimized for use in all Applied Biosystems™ real-time PCR instruments. And because it is a real-time PCR kit, no electrophoresis is required. In addition, the kit offers extremely high sensitivity, detecting targets with only 5 DNA copies.
The development and manufacture of this kit occurs through a partnership with Health in Code, S.L., which has a long history of GMO research and development.
Other related products include:
Taqman™ Roundup Ready Soya Quantification Kit
GMO Extraction Kit
TaqMan™ GMO Maize Quantification Kit
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
N.º de reacciones4 x 48 reactions
Línea de productosTaqMan
Tipo de productoGMO Screening Kit
Cantidad4 x 48 reactions
Tipo de muestraFood
ObjetivoDNA
Especificidad de dianaObjetivo específico
Método de detecciónSonda de cebado
GC-Rich PCR PerformanceHigh
Método de PCRqPCR
Unit SizeEach
Preguntas frecuentes
What is the TaqMan GMO Screening kit designed for?
The TaqMan GMO Screening kit is designed to perform four reactions for each sample analysed (P35S, TNOS, P34S-FMV, and plant).