TaqMan™ P35S Maize Quantification Kit
Thermo Scientific™

TaqMan™ P35S Maize Quantification Kit

The Taqman™ P35S Maize Quantification Kit is a food safety testing kit used to determine the number of copies ofRead more
Have Questions?
Catalog NumberQuantity
448197248 Reactions
Catalog number 4481972
Price (MXN)
-
Quantity:
48 Reactions
The Taqman™ P35S Maize Quantification Kit is a food safety testing kit used to determine the number of copies of P35S promoter present in genetically modified organisms compared to the number of copies of maize present in any food or feed sample. The benefits of the Taqman™ P35S Maize Quantification Kit over other commercially available products include:

• Efficiency—detection of small DNA fragments through use of Taqman™ MGB probes
• Sensitivity—quantification limit of 20 DNA copies; detection limit of 3 DNA copies (maize and P35S systems)
• Flexibility—adapted to processed DNAs

The Taqman™ P35S Maize Quantification Kit is effective in detecting the presence of genetically modified organisms through transgenic maize quantification, based on the detection of P35S promoter. The detection success of this promoter is due to this regulatory region being present in most of the transgenic events. In addition, the Taqman™ P35S Maize Quantification Kit contains all necessary reagents for quantification and has been optimized and validated for use with the Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument and the Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument.

The development and manufacture of this kit occurs through a partnership with Health in Code, S.L., which has a long history of GMO research and development.

Other related products include:
GMO Extraction Kit
TaqMan™ GMO Screening Kit
Taqman™ Roundup Ready Soya Quantification Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodPrimer-Probe Detection
No. of Reactions48 reactions
Quantity48 Reactions
Sample TypeFood
Target GeneP35S
Target MoleculeDNA
Target SpecificityTarget-Specific
Unit SizeEach