Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 2.5 μm - FAQs
Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 2.5 μm - FAQs
View additional product information for Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 2.5 μm - FAQs (A16501)
1 product FAQs found
What are Alignflow Flow Cytometry Alignment Beads used for?
Alignflow Flow Cytometry Alignment Beads are reliable references for aligning, focusing, and calibrating flow cytometers. The Alignflow beads are highly uniform with respect to size and fluorescence intensity, resulting in low CVs for both FSC and intensity. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material. They are available in 2.5 µM and 6.0 µM sizes for blue, red, and UV lasers. Alignflow Flow Cytometry Alignment Beads for UV Lasers are optimally excitable between 350-370 nm with emission in the range of 400-470 nm, but they can also be excited by a violet laser.